Search
Search

Ccdc136| Gene Symbol: Ccdc136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856382.3 | XP_004856439.2 | 2 - 3 | 198 | 56 | |
| XM_013076506.2 | XP_012931960.1 | 2 - 3 | 163 | 56 | ||
| XM_013076507.2 | XP_012931961.1 | 2 - 3 | 163 | 56 | ||
| XM_013076508.2 | XP_012931962.1 | 2 - 3 | 163 | 56 | ||
| XM_021237553.1 | XP_021093212.1 | 2 - 3 | 163 | 56 | ||
| XM_021237554.1 | XP_021093213.1 | 2 - 3 | 163 | 56 | ||
| XM_021237555.1 | XP_021093214.1 | 2 - 3 | 163 | 56 | ||
| XM_021237556.1 | XP_021093215.1 | 2 - 3 | 163 | 56 | ||
| XM_021237557.1 | XP_021093216.1 | 2 - 3 | 163 | 56 | ||
| XM_021237558.1 | XP_021093217.1 | 2 - 3 | 163 | 56 | ||
| XM_021237559.1 | XP_021093218.1 | 2 - 3 | 198 | 56 | ||
| XM_021237560.1 | XP_021093219.1 | 2 - 3 | 198 | 56 | ||
Set Membership: |
|