Search
Search

Snx30| Gene Symbol: Snx30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849563.2 | XP_004849620.1 | 5 - 6 | 914 | 67 | |
| XM_021262233.1 | XP_021117892.1 | 5 - 6 | 914 | 67 | ||
| XM_021262234.1 | XP_021117893.1 | 5 - 6 | 914 | 67 | ||
| XM_021262235.1 | XP_021117894.1 | 5 - 6 | 914 | 67 | ||
| XM_021262236.1 | XP_021117895.1 | 4 - 5 | 803 | 67 | ||
Set Membership: |
|