Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hg05229692_m1
          See other ABI3BP GE Assays ›
          Gene Symbol:
          Abi3bp
          Gene Name:
          ABI family member 3 binding protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.Un|NW_004624789: 14383162 - 14687395 on Build HetGla_female_1.0
          Species:
          Heterocephalus glaber
          Species Specific ID (Flybase ID):
          -
          Assay ID Hg05229692_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Abi3bp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_004857983.2 XP_004858040.1 3 - 4 415 67
          XM_013077097.2 XP_012932551.1 3 - 4 437 67
          XM_021238865.1 XP_021094524.1 3 - 4 415 67
          XM_021238866.1 XP_021094525.1 3 - 4 415 67
          XM_021238867.1 XP_021094526.1 3 - 4 415 67
          XM_021238869.1 XP_021094528.1 3 - 4 415 67
          XM_021238870.1 XP_021094529.1 3 - 4 415 67
          XM_021238871.1 XP_021094530.1 3 - 4 415 67
          XM_021238872.1 XP_021094531.1 3 - 4 415 67
          XM_021238873.1 XP_021094532.1 3 - 4 415 67
          XM_021238874.1 XP_021094533.1 3 - 4 415 67
          XM_021238875.1 XP_021094534.1 3 - 4 415 67
          XM_021238876.1 XP_021094535.1 3 - 4 415 67
          XM_021238877.1 XP_021094536.1 3 - 4 415 67
          XM_021238878.1 XP_021094537.1 3 - 4 415 67
          XM_021238880.1 XP_021094539.1 3 - 4 415 67
          XM_021238881.1 XP_021094540.1 3 - 4 415 67
          XM_021238882.1 XP_021094541.1 3 - 4 415 67
          XM_021238883.1 XP_021094542.1 3 - 4 415 67
          XM_021238884.1 XP_021094543.1 3 - 4 415 67
          XM_021238885.1 XP_021094544.1 3 - 4 415 67
          XM_021238886.1 XP_021094545.1 3 - 4 415 67
          XM_021238887.1 XP_021094546.1 3 - 4 415 67
          XM_021238888.1 XP_021094547.1 3 - 4 415 67
          XM_021238889.1 XP_021094548.1 3 - 4 415 67
          XM_021238890.1 XP_021094549.1 3 - 4 415 67
          XM_021238891.1 XP_021094550.1 3 - 4 415 67
          XM_021238892.1 XP_021094551.1 3 - 4 415 67
          XM_021238893.1 XP_021094552.1 3 - 4 415 67
          XM_021238894.1 XP_021094553.1 3 - 4 415 67
          XM_021238895.1 XP_021094554.1 3 - 4 415 67
          XM_021238896.1 XP_021094555.1 3 - 4 415 67
          XM_021238897.1 XP_021094556.1 3 - 4 415 67
          XM_021238898.1 XP_021094557.1 3 - 4 415 67
          XM_021238899.1 XP_021094558.1 3 - 4 415 67
          XM_021238900.1 XP_021094559.1 3 - 4 415 67
          XM_021238901.1 XP_021094560.1 3 - 4 415 67
          XM_021238902.1 XP_021094561.1 3 - 4 415 67
          XM_021238903.1 XP_021094562.1 3 - 4 415 67

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline