Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hg05232782_m1
See other ZNF446 GE Assays ›
Gene Symbol:
Znf446
Gene Name:
zinc finger protein 446
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624832: 97055 - 102643 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID
Hg05232782_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Znf446
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_004866530.3
XP_004866587.1
1 - 2
3418
109
XM_004866532.3
XP_004866589.1
1 - 2
3418
109
XM_004866534.3
XP_004866591.1
1 - 2
3418
109
XM_004866535.3
XP_004866592.1
1 - 2
3418
109
XM_004866537.3
XP_004866594.1
1 - 2
3418
109
XM_013066245.2
XP_012921699.1
1 - 2
3418
109
XM_013066249.2
XP_012921703.1
1 - 2
3418
109
XM_021246068.1
XP_021101727.1
1 - 2
3418
109
XM_021246069.1
XP_021101728.1
1 - 2
3418
109
XM_021246070.1
XP_021101729.1
1 - 2
3418
109
XM_021246071.1
XP_021101730.1
1 - 2
3418
109
XM_021246072.1
XP_021101731.1
1 - 2
3418
109
XM_021246073.1
XP_021101732.1
1 - 2
3418
109
XM_021246074.1
XP_021101733.1
1 - 2
3418
109
XM_021246076.1
XP_021101735.1
1 - 2
3418
109
XM_021246077.1
XP_021101736.1
1 - 2
3418
109
XM_021246078.1
XP_021101737.1
1 - 2
3418
109
XM_021246079.1
XP_021101738.1
1 - 2
3418
109
XM_021246080.1
XP_021101739.1
1 - 2
3418
109
XM_021246081.1
XP_021101740.1
1 - 2
3418
109
XM_021246083.1
XP_021101742.1
1 - 2
3418
109
XM_021246084.1
XP_021101743.1
1 - 2
3418
109
XM_021246085.1
XP_021101744.1
1 - 2
3418
109
XM_021246086.1
XP_021101745.1
1 - 2
3418
109
XM_021246087.1
XP_021101746.1
1 - 2
3418
109
XM_021246088.1
XP_021101747.1
1 - 2
3418
109
XM_021246089.1
XP_021101748.1
1 - 2
3418
109
XM_021246090.1
XP_021101749.1
1 - 2
3418
109
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
Probe spans exons
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline