Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hg05232784_m1
See other ZNF446 GE Assays ›
Gene Symbol:
Znf446
Gene Name:
zinc finger protein 446
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624832: 97055 - 102643 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID
Hg05232784_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Znf446
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_004866530.3
XP_004866587.1
3 - 4
3990
101
XM_004866532.3
XP_004866589.1
3 - 4
3990
101
XM_004866534.3
XP_004866591.1
3 - 4
3990
101
XM_004866535.3
XP_004866592.1
3 - 4
3990
101
XM_004866537.3
XP_004866594.1
3 - 4
3990
101
XM_013066245.2
XP_012921699.1
3 - 4
3990
101
XM_013066249.2
XP_012921703.1
3 - 4
3990
101
XM_021246068.1
XP_021101727.1
3 - 4
3990
101
XM_021246069.1
XP_021101728.1
3 - 4
3990
101
XM_021246070.1
XP_021101729.1
3 - 4
3990
101
XM_021246071.1
XP_021101730.1
3 - 4
3990
101
XM_021246072.1
XP_021101731.1
3 - 4
3990
101
XM_021246073.1
XP_021101732.1
3 - 4
3990
101
XM_021246074.1
XP_021101733.1
3 - 4
3990
101
XM_021246076.1
XP_021101735.1
3 - 4
3990
101
XM_021246077.1
XP_021101736.1
3 - 4
3990
101
XM_021246078.1
XP_021101737.1
3 - 4
3990
101
XM_021246079.1
XP_021101738.1
3 - 4
3990
101
XM_021246080.1
XP_021101739.1
3 - 4
3990
101
XM_021246081.1
XP_021101740.1
3 - 4
3990
101
XM_021246083.1
XP_021101742.1
3 - 4
3990
101
XM_021246084.1
XP_021101743.1
3 - 4
3990
101
XM_021246085.1
XP_021101744.1
3 - 4
3990
101
XM_021246086.1
XP_021101745.1
3 - 4
3990
101
XM_021246087.1
XP_021101746.1
3 - 4
3990
101
XM_021246088.1
XP_021101747.1
3 - 4
3990
101
XM_021246089.1
XP_021101748.1
3 - 4
3990
101
XM_021246090.1
XP_021101749.1
3 - 4
3990
101
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
3' Most
Probe spans exons
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
git-commit: c9e08c96761173abe34e68f880379696776a4827
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.44.1-2026.02.97.1.0