Search
Search

Erlec1| Gene Symbol: Erlec1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867291.3 | XP_004867348.1 | 1 - 2 | 372 | 86 | |
| XM_021246303.1 | XP_021101962.1 | 1 - 2 | 374 | 86 | ||
| XM_021246304.1 | XP_021101963.1 | 1 - 2 | 373 | 86 | ||
| XM_021246305.1 | XP_021101964.1 | 1 - 2 | 373 | 86 | ||
| XM_021246306.1 | XP_021101965.1 | 1 - 2 | 374 | 86 | ||
Set Membership: |
|