Search
Search

Cherp| Gene Symbol: Cherp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873271.2 | XP_004873328.1 | 4 - 5 | 636 | 61 | |
| XM_004873273.2 | XP_004873330.1 | 4 - 5 | 520 | 61 | ||
| XM_021251052.1 | XP_021106711.1 | 4 - 5 | 641 | 61 | ||
| XM_021251053.1 | XP_021106712.1 | 4 - 5 | 520 | 61 | ||
Set Membership: |
|