Search
Search

Fbxw8| Gene Symbol: Fbxw8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843712.3 | XP_004843769.1 | 5 - 6 | 811 | 61 | |
| XM_021257386.1 | XP_021113045.1 | 5 - 6 | 888 | 61 | ||
| XM_021257388.1 | XP_021113047.1 | 5 - 6 | 533 | 61 | ||
Set Membership: |
|