Search
Search

Cntn2| Gene Symbol: Cntn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862129.3 | XP_004862186.1 | 15 - 16 | 2432 | 68 | |
| XM_013078632.2 | XP_012934086.1 | 15 - 16 | 2354 | 68 | ||
| XM_021242458.1 | XP_021098117.1 | 15 - 16 | 2418 | 68 | ||
| XM_021242459.1 | XP_021098118.1 | 15 - 16 | 2461 | 68 | ||
Set Membership: |
|