Search
Search

Eps8l1| Gene Symbol: Eps8l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866685.3 | XP_004866742.2 | 3 - 4 | 370 | 62 | |
| XM_021246119.1 | XP_021101778.1 | 3 - 4 | 373 | 62 | ||
| XM_021246120.1 | XP_021101779.1 | 3 - 4 | 379 | 62 | ||
Set Membership: |
|