Search
Search

LOC101703341| Gene Symbol: LOC101703341 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864280.3 | XP_004864337.1 | 5 - 6 | 898 | 68 | |
| XM_013065325.2 | XP_012920779.1 | 5 - 6 | 898 | 68 | ||
Set Membership: |
|