Search
Search

Gnl1| Gene Symbol: Gnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847098.3 | XP_004847155.1 | 1 - 2 | 168 | 59 | |
| XM_004847099.3 | XP_004847156.1 | 1 - 2 | 168 | 59 | ||
| XM_004847100.3 | XP_004847157.1 | 1 - 2 | 168 | 59 | ||
Set Membership: |
|