Search
Search

Dsc3| Gene Symbol: Dsc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021265125.1 | XP_021120784.1 | 14 - 15 | 2503 | 65 | |
| XM_021265126.1 | XP_021120785.1 | 14 - 15 | 2503 | 65 | ||
Set Membership: |
|