Search
Search

Lrig1| Gene Symbol: Lrig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854171.3 | XP_004854228.1 | 7 - 8 | 1028 | 61 | |
| XM_004854172.3 | XP_004854229.1 | 7 - 8 | 1028 | 61 | ||
| XM_004854173.3 | XP_004854230.1 | 7 - 8 | 1028 | 61 | ||
| XM_021266005.1 | XP_021121664.1 | 7 - 8 | 1028 | 61 | ||
| XM_021266006.1 | XP_021121665.1 | 6 - 7 | 953 | 61 | ||
| XM_021266007.1 | XP_021121666.1 | 7 - 8 | 1028 | 61 | ||
| XM_021266008.1 | XP_021121667.1 | 5 - 6 | 881 | 61 | ||
Set Membership: |
|