Search
Search

Plxnb3| Gene Symbol: Plxnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013069362.2 | XP_012924816.1 | 18 - 19 | 3253 | 54 | |
| XM_013069363.2 | XP_012924817.1 | 18 - 19 | 3343 | 54 | ||
| XM_021252314.1 | XP_021107973.1 | 18 - 19 | 3416 | 54 | ||
| XM_021252315.1 | XP_021107974.1 | 18 - 19 | 3386 | 54 | ||
| XM_021252316.1 | XP_021107975.1 | 18 - 19 | 3416 | 54 | ||
| XM_021252317.1 | XP_021107976.1 | 18 - 19 | 3287 | 54 | ||
| XM_021252318.1 | XP_021107977.1 | 18 - 19 | 3390 | 54 | ||
| XM_021252320.1 | XP_021107979.1 | 18 - 19 | 3298 | 54 | ||
| XM_021252321.1 | XP_021107980.1 | 18 - 19 | 3416 | 54 | ||
Set Membership: |
|