Search
Search

Arhgef18| Gene Symbol: Arhgef18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021245189.1 | XP_021100848.1 | 22 - 23 | 3036 | 56 | |
| XM_021245190.1 | XP_021100849.1 | 22 - 23 | 2959 | 56 | ||
| XM_021245191.1 | XP_021100850.1 | 22 - 23 | 3031 | 56 | ||
Set Membership: |
|