Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00155582_m1
          See other MPPED2 GE Assays ›
          Gene Symbol:
          MPPED2
          Gene Name:
          metallophosphoesterase domain containing 2
          Gene Aliases:
          239FB, C11orf8
          Chromosome Location:
          Chr.11: 30384493 - 30586713 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00155582_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MPPED2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145399.1 NP_001138871.1 4 - 5 996 71
          NM_001584.2 NP_001575.1 3 - 4 653 71
          XM_005253110.3 XP_005253167.1 4 - 5 1326 71
          XM_005253111.2 XP_005253168.1 4 - 5 963 71
          XM_005253112.1 XP_005253169.1 4 - 5 791 71
          XM_011520346.1 XP_011518648.1 5 - 6 830 71
          XM_011520347.2 XP_011518649.1 4 - 5 1242 71
          XM_017018231.1 XP_016873720.1 5 - 6 812 71
          XM_017018232.1 XP_016873721.1 4 - 5 963 71
          XM_017018233.1 XP_016873722.1 5 - 6 1349 71
          XM_017018234.1 XP_016873723.1 5 - 6 1461 71
          XM_017018235.1 XP_016873724.1 5 - 6 1424 71
          XM_017018236.1 XP_016873725.1 3 - 4 1144 71
          XM_017018237.1 XP_016873726.1 3 - 4 724 71
          GenBank mRNA AB209163.1 - 4 - 5 894 71
          AK309809.1 - 5 - 6 830 71
          BC031582.1 - 3 - 4 669 5185528 71
          U57911.1 - 3 - 4 653 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100010440.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11001523.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MPPED2

          Panther Classification:

          Molecular Function -

          esterase

          Gene Ontology Categories:

          Function(s) Process(es)

          nervous system development
          protein binding
          hydrolase activity
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR11019019.hg.1 ENST00000358117.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11019019.hg.1 ENST00000448418.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11019019.hg.1 ENST00000524667.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11019019.hg.1 NM_001145399.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11019019.hg.1 NM_001584.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11019019.hg.1 TC11001523.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000358117.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000358117.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000358117.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000358117.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000358117.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000358117.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000358117.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000358117.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000358117.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000358117.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000358117.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000358117.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000358117.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000448418.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000448418.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000448418.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000448418.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000448418.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000448418.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000448418.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000448418.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000448418.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000448418.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000524636.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000524636.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000524636.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000524667.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000524667.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000524667.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000524667.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000524667.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000524667.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000524667.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000525519.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000525519.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000525519.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000526437.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000526437.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000526437.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000526437.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000526437.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000526437.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000526437.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000526437.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000526437.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000526437.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000526437.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000528686.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000528686.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000528686.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000528686.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000529220.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000529220.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000529220.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000529220.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000529220.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 ENST00000529220.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 NM_001145399.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 NM_001145399.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 NM_001145399.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 NM_001145399.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 NM_001145399.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 NM_001145399.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 NM_001145399.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 NM_001145399.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 NM_001145399.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 NM_001145399.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 NM_001145399.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 NM_001584.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 NM_001584.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 NM_001584.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 NM_001584.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 NM_001584.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 NM_001584.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 NM_001584.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 NM_001584.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 NM_001584.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 NM_001584.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 NM_001584.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 NM_001584.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010440.hg.1 NM_001584.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100179071.hg.1 ENST00000358117.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1100179071.hg.1 ENST00000448418.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100179071.hg.1 ENST00000524667.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100179071.hg.1 ENST00000526437.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100179071.hg.1 ENST00000529220.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100179071.hg.1 NM_001145399.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100179071.hg.1 NM_001584.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100179071.hg.1 ENST00000358117.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1100179071.hg.1 ENST00000448418.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100179071.hg.1 ENST00000524667.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100179071.hg.1 ENST00000526437.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100179071.hg.1 ENST00000529220.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100179071.hg.1 NM_001145399.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100179071.hg.1 NM_001584.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100179071.hg.1 TC1100010440.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000358117.9 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000358117.9 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000358117.9 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000358117.9 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000358117.9 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000358117.9 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000358117.9 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000358117.9 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000358117.9 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000358117.9 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000358117.9 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000358117.9 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000358117.9 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000448418.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000448418.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000448418.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000448418.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000448418.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000448418.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000448418.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000448418.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000448418.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000448418.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000524636.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000524636.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000524636.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000524667.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000524667.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000524667.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000524667.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000524667.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000524667.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000524667.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000525519.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000525519.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000525519.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000526437.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000526437.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000526437.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000526437.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000526437.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000526437.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000526437.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000526437.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000526437.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000526437.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000526437.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000528686.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000528686.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000528686.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000528686.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000529220.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000529220.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000529220.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000529220.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000529220.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 ENST00000529220.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 NM_001145399.1 Refseq
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 NM_001145399.1 Refseq
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 NM_001145399.1 Refseq
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 NM_001145399.1 Refseq
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 NM_001145399.1 Refseq
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 NM_001145399.1 Refseq
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 NM_001145399.1 Refseq
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 NM_001145399.1 Refseq
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 NM_001145399.1 Refseq
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 NM_001145399.1 Refseq
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 NM_001145399.1 Refseq
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 NM_001584.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 NM_001584.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 NM_001584.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 NM_001584.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 NM_001584.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 NM_001584.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 NM_001584.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 NM_001584.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 NM_001584.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 NM_001584.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 NM_001584.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 NM_001584.2 Refseq
          Affymetrix Human Clariom D Assay TC1100010440.hg.1 NM_001584.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline