Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00156454_m1
          See other CEBPG GE Assays ›
          Gene Symbol:
          CEBPG
          Gene Name:
          CCAAT/enhancer binding protein gamma
          Gene Aliases:
          GPE1BP, IG/EBP-1
          Chromosome Location:
          Chr.19: 33373669 - 33382686 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00156454_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CEBPG
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001806.3 NP_001797.1 1 - 2 226 131
          GenBank mRNA BC007582.1 - 1 - 2 565 3139979 131
          BC013128.1 - 1 - 2 144 3445301 131
          CB993585.1 - 219 131
          U20240.1 - 1 - 2 153 131

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900007777.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19000437.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the BC007582.1,BC013128.1,NM_001806.3,U20240.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CEBPG

          Panther Classification:

          Molecular Function -

          basic leucine zipper transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          liver development
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          immune response
          mRNA metabolic process
          B cell differentiation
          natural killer cell mediated cytotoxicity
          enucleate erythrocyte differentiation
          positive regulation of DNA binding
          negative regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of interferon-gamma biosynthetic process
          positive regulation of DNA repair
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of sequence-specific DNA binding transcription factor activity
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          protein binding
          transcription factor binding
          protein homodimerization activity
          sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding, bending
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR19005676.hg.1 ENST00000284000.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19005676.hg.1 NM_001806.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19005676.hg.1 TC19000437.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1900007777.hg.1 ENST00000284000.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007777.hg.1 ENST00000284000.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007777.hg.1 ENST00000284000.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007777.hg.1 ENST00000284000.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007777.hg.1 ENST00000284000.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007777.hg.1 ENST00000284000.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007777.hg.1 ENST00000284000.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007777.hg.1 ENST00000585933.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007777.hg.1 ENST00000585933.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007777.hg.1 ENST00000585933.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007777.hg.1 ENST00000585933.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007777.hg.1 ENST00000585933.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007777.hg.1 ENST00000585933.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007777.hg.1 NM_001252296.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007777.hg.1 NM_001252296.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007777.hg.1 NM_001252296.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007777.hg.1 NM_001252296.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007777.hg.1 NM_001252296.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007777.hg.1 NM_001806.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007777.hg.1 NM_001806.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007777.hg.1 NM_001806.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007777.hg.1 NM_001806.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007777.hg.1 NM_001806.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007777.hg.1 NM_001806.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19005675.hg.1 NM_001806.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19005675.hg.1 TC19000437.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19005682.hg.1 ENST00000284000.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19005682.hg.1 NM_001252296.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19005682.hg.1 NM_001806.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19005682.hg.1 TC19000437.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19005681.hg.1 ENST00000284000.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19005681.hg.1 NM_001252296.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19005681.hg.1 NM_001806.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19005681.hg.1 TC19000437.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19005677.hg.1 ENST00000284000.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19005677.hg.1 NM_001806.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19005677.hg.1 TC19000437.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19005680.hg.1 ENST00000284000.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19005680.hg.1 NM_001252296.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19005680.hg.1 NM_001806.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19005680.hg.1 TC19000437.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19005679.hg.1 ENST00000284000.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19005679.hg.1 NM_001252296.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19005679.hg.1 NM_001806.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19005679.hg.1 TC19000437.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1900157136.hg.1 ENST00000284000.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157136.hg.1 ENST00000585933.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157136.hg.1 NM_001252296.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900157136.hg.1 NM_001806.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900157136.hg.1 TC1900007777.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900157136.hg.1 ENST00000284000.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157136.hg.1 ENST00000585933.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157136.hg.1 NM_001252296.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900157136.hg.1 NM_001806.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900157137.hg.1 ENST00000284000.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157137.hg.1 ENST00000585933.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157137.hg.1 NM_001252296.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900157137.hg.1 NM_001806.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900157137.hg.1 TC1900007777.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900157137.hg.1 ENST00000284000.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157137.hg.1 ENST00000585933.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157137.hg.1 NM_001252296.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900157137.hg.1 NM_001806.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900157131.hg.1 ENST00000284000.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157131.hg.1 NM_001806.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900157131.hg.1 TC1900007777.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900157131.hg.1 ENST00000284000.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157131.hg.1 NM_001806.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900157132.hg.1 ENST00000284000.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157132.hg.1 NM_001806.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900157132.hg.1 TC1900007777.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900157132.hg.1 ENST00000284000.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157132.hg.1 NM_001806.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900157138.hg.1 ENST00000284000.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157138.hg.1 ENST00000585933.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157138.hg.1 NM_001252296.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900157138.hg.1 NM_001806.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900157138.hg.1 TC1900007777.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900157138.hg.1 ENST00000284000.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157138.hg.1 ENST00000585933.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157138.hg.1 NM_001252296.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900157138.hg.1 NM_001806.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900157130.hg.1 ENST00000284000.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157130.hg.1 TC1900007777.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900157130.hg.1 ENST00000284000.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900007777.hg.1 ENST00000284000.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900007777.hg.1 ENST00000284000.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900007777.hg.1 ENST00000284000.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900007777.hg.1 ENST00000284000.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900007777.hg.1 ENST00000284000.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900007777.hg.1 ENST00000284000.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900007777.hg.1 ENST00000284000.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900007777.hg.1 ENST00000585933.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900007777.hg.1 ENST00000585933.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900007777.hg.1 ENST00000585933.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900007777.hg.1 ENST00000585933.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900007777.hg.1 ENST00000585933.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900007777.hg.1 ENST00000585933.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900007777.hg.1 NM_001252296.1 Refseq
          Affymetrix Human Clariom D Assay TC1900007777.hg.1 NM_001252296.1 Refseq
          Affymetrix Human Clariom D Assay TC1900007777.hg.1 NM_001252296.1 Refseq
          Affymetrix Human Clariom D Assay TC1900007777.hg.1 NM_001252296.1 Refseq
          Affymetrix Human Clariom D Assay TC1900007777.hg.1 NM_001252296.1 Refseq
          Affymetrix Human Clariom D Assay TC1900007777.hg.1 NM_001806.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007777.hg.1 NM_001806.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007777.hg.1 NM_001806.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007777.hg.1 NM_001806.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007777.hg.1 NM_001806.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007777.hg.1 NM_001806.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900157135.hg.1 ENST00000284000.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157135.hg.1 ENST00000585933.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157135.hg.1 NM_001252296.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900157135.hg.1 NM_001806.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900157135.hg.1 TC1900007777.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900157135.hg.1 ENST00000284000.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157135.hg.1 ENST00000585933.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900157135.hg.1 NM_001252296.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900157135.hg.1 NM_001806.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline