Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006327.hg.1 |
TC09000571.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006327.hg.1 |
ENST00000259392.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006327.hg.1 |
ENST00000490809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006327.hg.1 |
NM_001860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006329.hg.1 |
TC09000571.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006329.hg.1 |
ENST00000259392.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006329.hg.1 |
ENST00000490809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006329.hg.1 |
NM_001860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006335.hg.1 |
TC09000571.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006335.hg.1 |
ENST00000259392.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006335.hg.1 |
NM_001860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006331.hg.1 |
TC09000571.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006331.hg.1 |
ENST00000259392.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006331.hg.1 |
NM_001860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006333.hg.1 |
TC09000571.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006333.hg.1 |
ENST00000259392.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006333.hg.1 |
NM_001860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008482.hg.1 |
ENST00000259392.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008482.hg.1 |
ENST00000259392.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008482.hg.1 |
ENST00000259392.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008482.hg.1 |
ENST00000259392.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008482.hg.1 |
ENST00000259392.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008482.hg.1 |
ENST00000259392.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008482.hg.1 |
ENST00000374220.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008482.hg.1 |
ENST00000374220.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008482.hg.1 |
ENST00000374220.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008482.hg.1 |
ENST00000374220.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008482.hg.1 |
ENST00000374220.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008482.hg.1 |
ENST00000374220.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008482.hg.1 |
ENST00000490809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008482.hg.1 |
ENST00000490809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008482.hg.1 |
ENST00000490809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008482.hg.1 |
NM_001860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008482.hg.1 |
NM_001860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008482.hg.1 |
NM_001860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008482.hg.1 |
NM_001860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008482.hg.1 |
NM_001860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008482.hg.1 |
NM_001860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156616.hg.1 |
ENST00000259392.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156616.hg.1 |
ENST00000374220.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156616.hg.1 |
NM_001860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156616.hg.1 |
ENST00000259392.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156616.hg.1 |
ENST00000374220.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156616.hg.1 |
NM_001860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156616.hg.1 |
TC0900008482.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900156603.hg.1 |
ENST00000259392.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156603.hg.1 |
ENST00000490809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156603.hg.1 |
NM_001860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156603.hg.1 |
ENST00000259392.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156603.hg.1 |
ENST00000490809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156603.hg.1 |
NM_001860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156603.hg.1 |
TC0900008482.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900156605.hg.1 |
ENST00000259392.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156605.hg.1 |
ENST00000374220.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156605.hg.1 |
ENST00000490809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156605.hg.1 |
NM_001860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156605.hg.1 |
ENST00000259392.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156605.hg.1 |
ENST00000374220.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156605.hg.1 |
ENST00000490809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156605.hg.1 |
NM_001860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156605.hg.1 |
TC0900008482.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900156614.hg.1 |
ENST00000259392.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156614.hg.1 |
ENST00000374220.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156614.hg.1 |
NM_001860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156614.hg.1 |
ENST00000259392.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156614.hg.1 |
ENST00000374220.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156614.hg.1 |
NM_001860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156614.hg.1 |
TC0900008482.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900008482.hg.1 |
ENST00000259392.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008482.hg.1 |
ENST00000259392.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008482.hg.1 |
ENST00000259392.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008482.hg.1 |
ENST00000259392.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008482.hg.1 |
ENST00000259392.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008482.hg.1 |
ENST00000259392.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008482.hg.1 |
ENST00000374220.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008482.hg.1 |
ENST00000374220.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008482.hg.1 |
ENST00000374220.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008482.hg.1 |
ENST00000374220.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008482.hg.1 |
ENST00000374220.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008482.hg.1 |
ENST00000374220.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008482.hg.1 |
ENST00000490809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008482.hg.1 |
ENST00000490809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008482.hg.1 |
ENST00000490809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008482.hg.1 |
NM_001860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008482.hg.1 |
NM_001860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008482.hg.1 |
NM_001860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008482.hg.1 |
NM_001860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008482.hg.1 |
NM_001860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008482.hg.1 |
NM_001860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156609.hg.1 |
ENST00000259392.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156609.hg.1 |
ENST00000374220.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156609.hg.1 |
NM_001860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156609.hg.1 |
ENST00000259392.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156609.hg.1 |
ENST00000374220.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156609.hg.1 |
NM_001860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156609.hg.1 |
TC0900008482.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900156612.hg.1 |
ENST00000259392.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156612.hg.1 |
ENST00000374220.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156612.hg.1 |
NM_001860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156612.hg.1 |
ENST00000259392.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156612.hg.1 |
ENST00000374220.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156612.hg.1 |
NM_001860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156612.hg.1 |
TC0900008482.hg.1 |
Clariom D TC |