| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19014409.hg.1 |
ENST00000309311.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19014409.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19014409.hg.1 |
TC19001054.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000384048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000384048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000594885.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000594885.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000594885.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000594885.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000596417.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000596417.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000596417.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000598182.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000598182.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000598182.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000598182.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000598182.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000598182.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000598436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000598436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000598436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000598436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000598436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000598436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000598436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000600720.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000600720.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000600720.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000600794.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000600794.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000600794.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
ENST00000600794.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NR_002602.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009316.hg.1 |
NR_002602.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900174382.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900174382.hg.1 |
ENST00000600794.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900174382.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900174382.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900174382.hg.1 |
ENST00000600794.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900174382.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900174382.hg.1 |
TC1900009316.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900174383.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900174383.hg.1 |
ENST00000596417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900174383.hg.1 |
ENST00000600794.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900174383.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900174383.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900174383.hg.1 |
ENST00000596417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900174383.hg.1 |
ENST00000600794.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900174383.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900174383.hg.1 |
TC1900009316.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000384048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000384048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000594885.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000594885.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000594885.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000594885.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000596417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000596417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000596417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000598182.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000598182.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000598182.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000598182.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000598182.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000598182.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000598436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000598436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000598436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000598436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000598436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000598436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000598436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000600720.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000600720.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000600720.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000600794.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000600794.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000600794.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
ENST00000600794.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NR_002602.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009316.hg.1 |
NR_002602.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900174384.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900174384.hg.1 |
ENST00000596417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900174384.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900174384.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900174384.hg.1 |
ENST00000596417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900174384.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900174384.hg.1 |
TC1900009316.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900174380.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900174380.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900174380.hg.1 |
ENST00000309311.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900174380.hg.1 |
NM_001961.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900174380.hg.1 |
TC1900009316.hg.1 |
Clariom D TC |