Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00160616_g1
          See other PSMB10 GE Assays ›
          Gene Symbol:
          PSMB10
          Gene Name:
          proteasome subunit beta 10
          Gene Aliases:
          LMP10, MECL1, beta2i
          Chromosome Location:
          Chr.16: 67934504 - 67936877 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00160616_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PSMB10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002801.3 NP_002792.1 2 - 3 275 75
          GenBank mRNA AK225658.1 - 2 - 3 248 75
          AK298012.1 - 1 - 2 472 75
          AK312208.1 - 2 - 3 235 75
          BC017198.1 - 2 - 3 226 3143189 75
          BC052369.1 - 2 - 3 213 4054165 75
          BM845639.1 - 2 - 3 248 75
          BP289065.1 - 2 - 3 275 75
          BT019723.1 - 2 - 3 147 75
          BT019724.1 - 2 - 3 147 75
          CR456982.1 - 2 - 3 147 75
          Y13640.1 - 2 - 3 235 75

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PSMB10

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          protein polyubiquitination
          cell morphogenesis
          stimulatory C-type lectin receptor signaling pathway
          antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
          regulation of cellular amino acid metabolic process
          humoral immune response
          viral process
          anaphase-promoting complex-dependent catabolic process
          tumor necrosis factor-mediated signaling pathway
          NIK/NF-kappaB signaling
          Fc-epsilon receptor signaling pathway
          T cell proliferation
          proteasome-mediated ubiquitin-dependent protein catabolic process
          regulation of mRNA stability
          T cell receptor signaling pathway
          negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
          positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
          Wnt signaling pathway, planar cell polarity pathway
          negative regulation of canonical Wnt signaling pathway
          positive regulation of canonical Wnt signaling pathway
          threonine-type endopeptidase activity
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000358514.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000358514.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000358514.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000358514.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000358514.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000358514.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000358514.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000358514.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000358514.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000358514.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000358514.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000570304.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000570304.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000570304.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000570304.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000570304.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000570304.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000570304.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000570304.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000570304.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000570304.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000570985.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000570985.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000570985.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000570985.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000570985.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000570985.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000574576.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000574576.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000574576.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000574576.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000574576.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000575556.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000575556.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000575556.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000575556.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000575556.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000575556.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000575556.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000575556.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 ENST00000575556.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 NM_002801.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 NM_002801.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 NM_002801.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 NM_002801.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 NM_002801.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 NM_002801.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 NM_002801.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 NM_002801.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 NM_002801.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600011553.hg.1 NM_002801.3 Refseq
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000358514.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000358514.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000358514.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000358514.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000358514.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000358514.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000358514.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000358514.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000358514.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000358514.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000358514.8 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000570304.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000570304.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000570304.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000570304.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000570304.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000570304.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000570304.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000570304.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000570304.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000570304.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000570985.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000570985.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000570985.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000570985.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000570985.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000570985.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000574576.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000574576.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000574576.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000574576.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000574576.2 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000575556.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000575556.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000575556.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000575556.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000575556.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000575556.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000575556.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000575556.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 ENST00000575556.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 NM_002801.3 Refseq
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 NM_002801.3 Refseq
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 NM_002801.3 Refseq
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 NM_002801.3 Refseq
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 NM_002801.3 Refseq
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 NM_002801.3 Refseq
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 NM_002801.3 Refseq
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 NM_002801.3 Refseq
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 NM_002801.3 Refseq
          Affymetrix Human Clariom D Assay TC1600011553.hg.1 NM_002801.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline