Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00167927_m1
          See other CYP1A2 GE Assays ›
          Gene Symbol:
          CYP1A2
          Gene Name:
          cytochrome P450 family 1 subfamily A member 2
          Gene Aliases:
          CP12, CYPIA2, P3-450, P450(PA)
          Chromosome Location:
          Chr.15: 74748845 - 74756607 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00167927_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (6)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CYP1A2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000761.5 NP_000752.2 2 - 3 893 67
          GenBank mRNA AF182274.1 - 1 - 2 839 67
          BC067424.1 - 2 - 3 875 67
          BC067425.1 - 2 - 3 875 67
          BC067426.1 - 2 - 3 875 67
          BC067427.1 - 2 - 3 875 67
          BC067428.1 - 2 - 3 875 67
          BC067429.1 - 2 - 3 875 67
          M55053.1 - 1 - 2 839 67
          Z00036.1 - 1 - 2 839 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1500007855.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC15000684.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection P450 Recommended Assay Homo sapiens Human CYP450 and other Oxygenases Human Immune Response Inventoried MicroArray Quality Control (MAQC) Human_Immune Human CYP450 + oxygenases Human_CYP450_and_other_Oxygenases Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CYP1A2

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          fatty acid metabolic process
          steroid catabolic process
          porphyrin-containing compound metabolic process
          xenobiotic metabolic process
          steroid metabolic process
          cholesterol metabolic process
          estrogen metabolic process
          toxin biosynthetic process
          toxin metabolic process
          post-embryonic development
          alkaloid metabolic process
          regulation of gene expression
          monoterpenoid metabolic process
          dibenzo-p-dioxin metabolic process
          arachidonic acid metabolic process
          epoxygenase P450 pathway
          lung development
          methylation
          monocarboxylic acid metabolic process
          xenobiotic catabolic process
          hormone metabolic process
          retinol metabolic process
          long-chain fatty acid biosynthetic process
          cellular respiration
          aflatoxin metabolic process
          hydrogen peroxide biosynthetic process
          oxidative demethylation
          cellular response to cadmium ion
          omega-hydroxylase P450 pathway
          olefinic compound metabolic process
          monooxygenase activity
          iron ion binding
          protein binding
          steroid hydroxylase activity
          electron carrier activity
          oxidoreductase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen
          lyase activity
          enzyme binding
          heme binding
          demethylase activity
          caffeine oxidase activity
          metal ion binding
          estrogen 16-alpha-hydroxylase activity
          estrogen 2-hydroxylase activity
          hydroperoxy icosatetraenoate dehydratase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1500007855.hg.1 ENST00000343932.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007855.hg.1 ENST00000343932.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007855.hg.1 ENST00000343932.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007855.hg.1 ENST00000343932.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007855.hg.1 ENST00000343932.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007855.hg.1 ENST00000343932.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007855.hg.1 ENST00000343932.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007855.hg.1 ENST00000343932.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007855.hg.1 NM_000761.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007855.hg.1 NM_000761.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007855.hg.1 NM_000761.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007855.hg.1 NM_000761.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007855.hg.1 NM_000761.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007855.hg.1 NM_000761.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007855.hg.1 NM_000761.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007855.hg.1 NM_000761.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007855.hg.1 NM_000761.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15006009.hg.1 ENST00000343932.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15006009.hg.1 NM_000761.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15006009.hg.1 TC15000684.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1500156032.hg.1 ENST00000343932.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1500156032.hg.1 NM_000761.4 Refseq
          Affymetrix Human Clariom D Assay PSR1500156032.hg.1 ENST00000343932.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1500156032.hg.1 NM_000761.4 Refseq
          Affymetrix Human Clariom D Assay PSR1500156032.hg.1 TC1500007855.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1500007855.hg.1 ENST00000343932.4 Ensembl
          Affymetrix Human Clariom D Assay TC1500007855.hg.1 ENST00000343932.4 Ensembl
          Affymetrix Human Clariom D Assay TC1500007855.hg.1 ENST00000343932.4 Ensembl
          Affymetrix Human Clariom D Assay TC1500007855.hg.1 ENST00000343932.4 Ensembl
          Affymetrix Human Clariom D Assay TC1500007855.hg.1 ENST00000343932.4 Ensembl
          Affymetrix Human Clariom D Assay TC1500007855.hg.1 ENST00000343932.4 Ensembl
          Affymetrix Human Clariom D Assay TC1500007855.hg.1 ENST00000343932.4 Ensembl
          Affymetrix Human Clariom D Assay TC1500007855.hg.1 ENST00000343932.4 Ensembl
          Affymetrix Human Clariom D Assay TC1500007855.hg.1 NM_000761.4 Refseq
          Affymetrix Human Clariom D Assay TC1500007855.hg.1 NM_000761.4 Refseq
          Affymetrix Human Clariom D Assay TC1500007855.hg.1 NM_000761.4 Refseq
          Affymetrix Human Clariom D Assay TC1500007855.hg.1 NM_000761.4 Refseq
          Affymetrix Human Clariom D Assay TC1500007855.hg.1 NM_000761.4 Refseq
          Affymetrix Human Clariom D Assay TC1500007855.hg.1 NM_000761.4 Refseq
          Affymetrix Human Clariom D Assay TC1500007855.hg.1 NM_000761.4 Refseq
          Affymetrix Human Clariom D Assay TC1500007855.hg.1 NM_000761.4 Refseq
          Affymetrix Human Clariom D Assay TC1500007855.hg.1 NM_000761.4 Refseq

          Back To Top
           

          Citations (6)

          Aqueous extract from Spanish black radish (Raphanus sativus L. Var. niger) induces detoxification enzymes in the HepG2 human hepatoma cell line.
          Authors
          Hanlon PR, Webber DM, Barnes DM
          Journal
          J Agric Food Chem - Page No 6439 (Wed Aug 08 00:00:00 UTC 2007)
          Volume (Year)
          55 (2007)
          Product Usage
          -
          Disturbed expression of phase I and phase II estrogen-metabolizing enzymes in endometrial cancer: lower levels of CYP1B1 and increased expression of S-COMT.
          Authors
          Hevir N, Sinkovec J, Rizner TL
          Journal
          Mol Cell Endocrinol - Page No 158 (Sat Jan 01 00:00:00 UTC 2011)
          Volume (Year)
          331 (2011)
          Product Usage
          -
          Development of stably transfected human and rat hepatoma cell lines for the species-specific assessment of xenobiotic response enhancer module (XREM)-dependent induction of drug metabolism.
          Authors
          Fery Y, Mueller SO, Schrenk D
          Journal
          Toxicology - Page No 11 (Tue Nov 09 00:00:00 UTC 2010)
          Volume (Year)
          277 (2010)
          Product Usage
          -
          The use of human hepatocytes to investigate drug metabolism and CYP enzyme induction.
          Authors
          Klieber S, Torreilles F, Guillou F, Fabre G
          Journal
          Methods Mol Biol - Page No 295 (Fri Jan 01 00:00:00 UTC 2010)
          Volume (Year)
          640 (2010)
          Product Usage
          -
          Robust expansion of human hepatocytes in Fah-/-/Rag2-/-/Il2rg-/- mice.
          Authors
          Azuma H, Paulk N, Ranade A, Dorrell C, Al-Dhalimy M, Ellis E, Strom S, Kay MA, Finegold M, Grompe M
          Journal
          Nat Biotechnol - Page No 903 (Wed Aug 01 00:00:00 UTC 2007)
          Volume (Year)
          25 (2007)
          Product Usage
          -
          Simultaneous assessment of cytochrome P450 activity in cultured human hepatocytes for compound-mediated induction of CYP3A4, CYP2B6, and CYP1A2.
          Authors
          Rhodes SP, Otten JN, Hingorani GP, Hartley DP, Franklin RB
          Journal
          J Pharmacol Toxicol Methods - Page No (Wed Nov 24 00:00:00 UTC 2010)
          Volume (Year)
           (2010)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline