Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00169777_m1
          See other PECAM1 GE Assays ›
          Gene Symbol:
          PECAM1
          Gene Name:
          platelet and endothelial cell adhesion molecule 1
          Gene Aliases:
          CD31, CD31/EndoCAM, GPIIA', PECA1, PECAM-1, endoCAM
          Chromosome Location:
          Chr.17: 64319415 - 64413844 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00169777_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PECAM1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000442.4 NP_000433.4 14 - 15 2376 65
          XM_005276882.1 XP_005276939.1 13 - 14 2599 65
          XM_011524889.2 XP_011523191.1 15 - 16 2392 65
          XM_011524890.1 XP_011523192.1 15 - 16 2341 65
          XM_017024738.1 XP_016880227.1 15 - 16 2285 65
          XM_017024739.1 XP_016880228.1 13 - 14 2608 65
          GenBank mRNA AF281287.1 - 14 - 15 2246 65
          AF281288.1 - 14 - 15 2246 65
          AF281289.1 - 14 - 15 2246 65
          AF281290.1 - 14 - 15 2246 65
          AF281291.1 - 14 - 15 2246 65
          AF281292.1 - 14 - 15 2246 65
          AF281293.1 - 14 - 15 2246 65
          AF281294.1 - 14 - 15 2246 65
          AF281295.1 - 14 - 15 2246 65
          AF281296.1 - 14 - 15 2246 65
          AF281297.1 - 14 - 15 2246 65
          AF281298.1 - 14 - 15 2246 65
          AF281299.1 - 14 - 15 2246 65
          AF281300.1 - 14 - 15 2246 65
          AF281301.1 - 14 - 15 2246 65
          AF393676.1 - 14 - 15 2246 65
          AF393677.1 - 14 - 15 2246 65
          AF393678.1 - 14 - 15 2246 65
          AK290692.1 - 14 - 15 2179 65
          AK303959.1 - 9 - 10 1150 65
          AK304166.1 - 13 - 14 2176 65
          BC022512.1 - 14 - 15 2356 4816878 65
          BC051822.1 - 14 - 15 2376 5768579 65
          JQ287500.1 - 14 - 15 2163 65
          M28526.1 - 14 - 15 2304 65
          M37780.1 - 14 - 15 2288 65

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Immune Tolerance Network Mammalian Gene Collection Homo sapiens Human Tumor Metastasis Panel Human Stem Cell Pluripotency Human Extracellular Matrix and Adhesion Molecules Human Hematopoeisis Human Signal Transduction Pathways Human Angiogenesis Human Cell Surface Markers Inventoried Human_Angiogenesis Human_Cell_Surface_Markers Human Tumor Metastasis Human_Hematopoisis Human_Signal_Transduction_Pathways Human_Stem_Cell Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PECAM1

          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          platelet degranulation
          phagocytosis
          cell adhesion
          signal transduction
          cell recognition
          extracellular matrix organization
          leukocyte migration
          diapedesis
          glomerular endothelium development
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000561739.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000561739.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000561739.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000563523.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000563523.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000563523.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000563523.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000564866.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000564866.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000564866.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000564866.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000564866.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000564866.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000564866.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000566422.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000566422.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000566422.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000566422.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000566616.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000566616.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000566616.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000568702.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000568702.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000568702.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000568702.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000568702.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000569530.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000569530.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000569530.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000569967.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000569967.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000569967.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000569967.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 ENST00000569967.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000561739.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000561739.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000561739.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000563523.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000563523.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000563523.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000563523.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000563924.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000564866.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000564866.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000564866.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000564866.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000564866.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000564866.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000564866.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000566422.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000566422.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000566422.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000566422.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000566616.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000566616.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000566616.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000568702.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000568702.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000568702.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000568702.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000568702.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000569530.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000569530.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000569530.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000569967.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000569967.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000569967.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000569967.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 ENST00000569967.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 NM_000442.4 Refseq
          Affymetrix Human Clariom D Assay TC1700011442.hg.1 NM_000442.4 Refseq

          Back To Top
           

          Citations (1)

          Functional inhibition of NF-kappaB signal transduction in alphavbeta3 integrin expressing endothelial cells by using RGD-PEG-modified adenovirus with a mutant IkappaB gene.
          Authors
          Ogawara K, Kuldo JM, Oosterhuis K, Kroesen BJ, Rots MG, Trautwein C, Kimura T, Haisma HJ, Molema G
          Journal
          Arthritis Res Ther - Page No R32 (Sun Jan 01 00:00:00 UTC 2006)
          Volume (Year)
          8 (2006)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline