| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000392029.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000392029.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000392029.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000392029.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000392029.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000392029.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000392029.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000392029.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000443059.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
ENST00000443059.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010798.hg.1 |
NR_026900.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01024124.hg.1 |
ENST00000367600.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01024124.hg.1 |
ENST00000367602.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01024124.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01024124.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01024124.hg.1 |
TC01001559.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000392029.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000392029.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000392029.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000392029.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000392029.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000392029.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000392029.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000392029.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000443059.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
ENST00000443059.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010798.hg.1 |
NR_026900.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100179346.hg.1 |
ENST00000367600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100179346.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100179346.hg.1 |
ENST00000392029.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100179346.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100179346.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100179346.hg.1 |
ENST00000367600.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100179346.hg.1 |
ENST00000367602.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100179346.hg.1 |
ENST00000392029.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100179346.hg.1 |
NM_001004128.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100179346.hg.1 |
NM_002826.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100179346.hg.1 |
TC0100010798.hg.1 |
Clariom D TC |