Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00171022_m1
          See other CXCL12 GE Assays ›
          Gene Symbol:
          CXCL12
          Gene Name:
          C-X-C motif chemokine ligand 12
          Gene Aliases:
          IRH, PBSF, SCYB12, SDF1, TLSF, TPAR1
          Chromosome Location:
          Chr.10: 44370165 - 44385097 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00171022_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CXCL12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000609.7 NP_000600.1 2 - 3 273 77
          NM_001033886.2 NP_001029058.1 2 - 3 273 77
          NM_001178134.2 NP_001171605.1 2 - 3 273 77
          NM_199168.4 NP_954637.1 2 - 3 273 77
          GenBank mRNA AK292628.1 - 2 - 3 276 77
          AK311814.1 - 2 - 3 271 77
          AY644456.1 - 2 - 3 230 77
          BC031072.1 - 2 - 3 265 77
          BC039893.1 - 2 - 3 251 77
          CD172464.1 - 2 - 3 271 77
          CR450283.1 - 2 - 3 181 77
          DA266752.1 - 2 - 3 274 77
          DQ345517.1 - 2 - 3 225 77
          DQ345518.1 - 2 - 3 220 77
          DQ345519.1 - 2 - 3 225 77
          DQ345520.1 - 2 - 3 225 77
          L36033.1 - 2 - 3 260 77
          L36034.1 - 2 - 3 260 77
          U16752.1 - 2 - 3 261 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000010468.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10001212.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Immune Tolerance Network Mammalian Gene Collection Homo sapiens Human Chemokine Panel Human Tumor Metastasis Panel Human Angiogenesis HUMAN 14-3-3 INDUCED INTRACELLULAR SIGNALING HUMAN P53 MEDIATED APOPTOSIS HUMAN IL-6 PATHWAY Inventoried Human_Angiogenesis Human_Chemokine Human Tumor Metastasis Human_IL-6_Pathway Human_14-3-3_Induced_Intracellular_Signaling Human_p53_Mediated_Apoptosis Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CXCL12

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          neuron migration
          positive regulation of endothelial cell proliferation
          cellular calcium ion homeostasis
          chemotaxis
          defense response
          immune response
          cell adhesion
          signal transduction
          G-protein coupled receptor signaling pathway
          adenylate cyclase-activating G-protein coupled receptor signaling pathway
          adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
          axon guidance
          blood circulation
          regulation of actin polymerization or depolymerization
          positive regulation of cell proliferation
          adult locomotory behavior
          response to mechanical stimulus
          response to virus
          T cell chemotaxis
          telencephalon cell migration
          positive regulation of cell migration
          organ regeneration
          killing of cells of other organism
          positive regulation of dopamine secretion
          integrin activation
          chemokine (C-X-C motif) ligand 12 signaling pathway
          CXCL12-activated CXCR4 signaling pathway
          response to peptide hormone
          positive regulation of neuron differentiation
          positive regulation of cell adhesion
          positive regulation of axon extension involved in axon guidance
          positive regulation of chemotaxis
          induction of positive chemotaxis
          detection of temperature stimulus involved in sensory perception of pain
          detection of mechanical stimulus involved in sensory perception of pain
          regulation of calcium ion transport
          cell chemotaxis
          antimicrobial humoral immune response mediated by antimicrobial peptide
          chemokine-mediated signaling pathway
          positive regulation of monocyte chemotaxis
          positive regulation of calcium ion import
          negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
          negative regulation of leukocyte tethering or rolling
          positive regulation of vasculature development
          response to ultrasound
          cellular response to chemokine
          positive regulation of T cell migration
          negative regulation of dendritic cell apoptotic process
          receptor binding
          cytokine activity
          integrin binding
          protein binding
          chemokine activity
          growth factor activity
          chemokine receptor binding
          CXCR chemokine receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000343575.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000343575.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000343575.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000343575.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000343575.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000343575.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000343575.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000343575.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000343575.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000343575.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374426.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374426.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374426.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374426.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374426.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374426.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374426.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374429.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374429.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374429.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374429.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374429.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374429.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374429.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374429.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374429.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395793.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395793.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395793.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395793.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395793.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395794.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395794.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395794.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395794.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395794.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395794.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395794.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395795.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395795.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395795.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395795.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000488591.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000488591.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000488591.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000488591.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000488591.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000496375.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000496375.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000496375.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000496375.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000496375.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_000609.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_000609.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_000609.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_000609.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_000609.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_000609.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_000609.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_000609.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_000609.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001033886.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001033886.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001033886.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001033886.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001033886.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001033886.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001033886.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001178134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001178134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001178134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001178134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001178134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001178134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001178134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001178134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001178134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001178134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001277990.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001277990.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001277990.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001277990.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001277990.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001277990.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_199168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_199168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_199168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_199168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_199168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_199168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_199168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_199168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_199168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_199168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_199168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_199168.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline