Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00171042_m1
          See other CXCL10 GE Assays ›
          Gene Symbol:
          CXCL10
          Gene Name:
          C-X-C motif chemokine ligand 10
          Gene Aliases:
          C7, IFI10, INP10, IP-10, SCYB10, crg-2, gIP-10, mob-1
          Chromosome Location:
          Chr.4: 76021116 - 76023536 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00171042_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (17)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CXCL10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001565.3 NP_001556.2 1 - 2 168 98
          GenBank mRNA BC010954.1 - 1 - 2 133 4274617 98
          X02530.1 - 1 - 2 129 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0400011053.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC04001305.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Immune Tolerance Network Mammalian Gene Collection Recommended Assay Homo sapiens Human Chemokine Panel Human Immune Response Human Angiogenesis Inventoried Human_Immune Human_Angiogenesis Human_Chemokine Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CXCL10

          Panther Classification:

          Molecular Function -

          chemokine

          Gene Ontology Categories:

          Function(s) Process(es)

          chemotaxis
          inflammatory response
          immune response
          signal transduction
          cell surface receptor signaling pathway
          G-protein coupled receptor signaling pathway
          cell-cell signaling
          muscle organ development
          blood circulation
          positive regulation of cell proliferation
          response to cold
          response to gamma radiation
          T cell chemotaxis
          regulation of T cell chemotaxis
          response to auditory stimulus
          negative regulation of angiogenesis
          positive regulation of cAMP metabolic process
          response to vitamin D
          cellular response to heat
          endothelial cell activation
          regulation of cell proliferation
          positive regulation of cAMP-mediated signaling
          negative regulation of myoblast differentiation
          regulation of protein kinase activity
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of release of sequestered calcium ion into cytosol
          defense response to virus
          chemokine-mediated signaling pathway
          cellular response to lipopolysaccharide
          positive regulation of monocyte chemotaxis
          regulation of endothelial tube morphogenesis
          negative regulation of myoblast fusion
          positive regulation of T cell migration
          receptor binding
          protein binding
          chemokine activity
          heparin binding
          cAMP-dependent protein kinase regulator activity
          CXCR3 chemokine receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR04017887.hg.1 ENST00000306602.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04017887.hg.1 NM_001565.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04017887.hg.1 TC04001305.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0400011053.hg.1 ENST00000306602.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011053.hg.1 ENST00000306602.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011053.hg.1 ENST00000306602.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011053.hg.1 ENST00000306602.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011053.hg.1 ENST00000306602.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011053.hg.1 NM_001565.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011053.hg.1 NM_001565.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011053.hg.1 NM_001565.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011053.hg.1 NM_001565.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011053.hg.1 NM_001565.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011053.hg.1 NM_001565.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400175301.hg.1 ENST00000306602.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400175301.hg.1 NM_001565.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400175301.hg.1 ENST00000306602.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400175301.hg.1 NM_001565.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400175301.hg.1 TC0400011053.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0400011053.hg.1 ENST00000306602.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011053.hg.1 ENST00000306602.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011053.hg.1 ENST00000306602.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011053.hg.1 ENST00000306602.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011053.hg.1 ENST00000306602.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011053.hg.1 NM_001565.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011053.hg.1 NM_001565.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011053.hg.1 NM_001565.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011053.hg.1 NM_001565.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011053.hg.1 NM_001565.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011053.hg.1 NM_001565.3 Refseq

          Back To Top
           

          Citations (17)

          The synthetic liver X receptor agonist GW3965 reduces tissue factor production and inflammatory responses in human islets in vitro.
          Authors
          Scholz H, Lund T, Dahle MK, Collins JL, Korsgren O, Wang JE, Foss A
          Journal
          Diabetologia - Page No 1352 (Wed Jul 01 00:00:00 UTC 2009)
          Volume (Year)
          52 (2009)
          Product Usage
          -
          Intrahepatic levels of CXCR3-associated chemokines correlate with liver inflammation and fibrosis in chronic hepatitis C.
          Authors
          Zeremski M, Petrovic LM, Chiriboga L, Brown QB, Yee HT, Kinkhabwala M, Jacobson IM, Dimova R, Markatou M, Talal AH
          Journal
          Hepatology - Page No 1440 (Sat Nov 01 00:00:00 UTC 2008)
          Volume (Year)
          48 (2008)
          Product Usage
          -
          No evidence of altered alveolar macrophage polarization, but reduced expression of TLR2, in bronchoalveolar lavage cells in sarcoidosis.
          Authors
          Wiken M, Idali F, Al Hayja MA, Grunewald J, Eklund A, Wahlstrom J
          Journal
          Respir Res - Page No 121 (Fri Jan 01 00:00:00 UTC 2010)
          Volume (Year)
          11 (2010)
          Product Usage
          -
          Gene expression profiles in psoriasis: analysis of impact of body site location and clinical severity.
          Authors
          Quekenborn-Trinquet V, Fogel P, Aldana-Jammayrac O, Ancian P, Demarchez M, Rossio P, Richards HL, Kirby B, Nguyen C, Voegel JJ, Griffiths CE
          Journal
          Br J Dermatol - Page No 489 (Tue Mar 01 00:00:00 UTC 2005)
          Volume (Year)
          152 (2005)
          Product Usage
          -
          Gene-expression analysis of polyI:C-stimulated primary human conjunctival epithelial cells.
          Authors
          Ueta M, Mizushima K, Yokoi N, Naito Y, Kinoshita S
          Journal
          Br J Ophthalmol - Page No 1528 (Mon Nov 01 00:00:00 UTC 2010)
          Volume (Year)
          94 (2010)
          Product Usage
          -
          Inhibition of dynamin-dependent endocytosis interferes with type III IFN expression in bacteria-infected human monocyte-derived DCs.
          Authors
          Pietila TE, Latvala S, Osterlund P, Julkunen I
          Journal
          J Leukoc Biol - Page No 665 (Fri Oct 01 00:00:00 UTC 2010)
          Volume (Year)
          88 (2010)
          Product Usage
          -
          Evaluation of CXCL9 and CXCL10 as circulating biomarkers of human cardiac allograft rejection.
          Authors
          Karason K, Jernas M, Hagg DA, Svensson PA
          Journal
          Bmc Cardiovasc Disord - Page No 29 (Sun Jan 01 00:00:00 UTC 2006)
          Volume (Year)
          6 (2006)
          Product Usage
          -
          Hepatitis C virus proteins interfere with the activation of chemokine gene promoters and downregulate chemokine gene expression.
          Authors
          Sillanpaa M, Kaukinen P, Melen K, Julkunen I
          Journal
          J Gen Virol - Page No 432 (Fri Feb 01 00:00:00 UTC 2008)
          Volume (Year)
          89 (2008)
          Product Usage
          -
          Identification of new sensitive biomarkers for the in vivo response to interferon-beta treatment in multiple sclerosis using DNA-array evaluation.
          Authors
          Sellebjerg F, Krakauer M, Hesse D, Ryder LP, Alsing I, Jensen PE, Koch-Henriksen N, Svejgaard A, Soelberg Sorensen P
          Journal
          Eur J Neurol - Page No 1291 (Tue Dec 01 00:00:00 UTC 2009)
          Volume (Year)
          16 (2009)
          Product Usage
          -
          15-Lipoxygenase-1 induces expression and release of chemokines in cultured human lung epithelial cells.
          Authors
          Liu C, Xu D, Liu L, Schain F, Brunnstrom A, Bjorkholm M, Claesson HE, Sjoberg J
          Journal
          Am J Physiol Lung Cell Mol Physiol - Page No L196 (Wed Jul 01 00:00:00 UTC 2009)
          Volume (Year)
          297 (2009)
          Product Usage
          -
          STAT1 signaling is associated with acquired crossresistance to doxorubicin and radiation in myeloma cell lines.
          Authors
          Fryknas M, Dhar S, Oberg F, Rickardson L, Rydaker M, Goransson H, Gustafsson M, Pettersson U, Nygren P, Larsson R, Isaksson A
          Journal
          Int J Cancer - Page No 189 (Mon Jan 01 00:00:00 UTC 2007)
          Volume (Year)
          120 (2007)
          Product Usage
          -
          Plasmacytoid dendritic cells initiate a complex chemokine and cytokine network and are a viable drug target in chronic HCV patients.
          Authors
          Decalf J, Fernandes S, Longman R, Ahloulay M, Audat F, Lefrerre F, Rice CM, Pol S, Albert ML
          Journal
          J Exp Med - Page No 2423 (Mon Oct 01 00:00:00 UTC 2007)
          Volume (Year)
          204 (2007)
          Product Usage
          -
          Using distinct molecular signatures of human monocytes and dendritic cells to predict adjuvant activity and pyrogenicity of TLR agonists.
          Authors
          Kamgang RK, Ramos I, Rodrigues Duarte L, Ghielmetti M, Freudenberg M, Dahinden C, Padovan E
          Journal
          Med Microbiol Immunol - Page No 369 (Mon Dec 01 00:00:00 UTC 2008)
          Volume (Year)
          197 (2008)
          Product Usage
          -
          cDNA microarray analysis of serially sampled cervical cancer specimens from patients treated with thermochemoradiotherapy.
          Authors
          Borkamo ED, Schem BC, Fluge O, Bruland O, Dahl O, Mella O
          Journal
          Int J Radiat Oncol Biol Phys - Page No 1562 (Tue Dec 01 00:00:00 UTC 2009)
          Volume (Year)
          75 (2009)
          Product Usage
          -
          Adenovirus-directed ocular innate immunity: the role of conjunctival defensin-like chemokines (IP-10, I-TAC) and phagocytic human defensin-alpha.
          Authors
          Harvey SA, Romanowski EG, Yates KA, Gordon YJ
          Journal
          Invest Ophthalmol Vis Sci - Page No 3657 (Sat Oct 01 00:00:00 UTC 2005)
          Volume (Year)
          46 (2005)
          Product Usage
          -
          Differential expression of immunoregulatory genes in monocytes in response to Porphyromonas gingivalis and Escherichia coli lipopolysaccharide.
          Authors
          Barksby HE, Nile CJ, Jaedicke KM, Taylor JJ, Preshaw PM
          Journal
          Clin Exp Immunol - Page No 479 (Mon Jun 01 00:00:00 UTC 2009)
          Volume (Year)
          156 (2009)
          Product Usage
          -
          Chemokine expression in melanoma metastases associated with CD8+ T-cell recruitment.
          Authors
          Harlin H, Meng Y, Peterson AC, Zha Y, Tretiakova M, Slingluff C, McKee M, Gajewski TF
          Journal
          Cancer Res - Page No 3077 (Wed Apr 01 00:00:00 UTC 2009)
          Volume (Year)
          69 (2009)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-hrdnk:80/100.66.78.145:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0