Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00174114_m1
          See other IL2 GE Assays ›
          Gene Symbol:
          IL2
          Gene Name:
          interleukin 2
          Gene Aliases:
          IL-2, TCGF, lymphokine
          Chromosome Location:
          Chr.4: 122449479 - 122456495 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00174114_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (18)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: IL2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000586.3 NP_000577.2 2 - 3 267 94
          XM_017008177.1 XP_016863666.1 2 - 3 233 94
          GenBank mRNA A14844.1 - 2 - 3 222 94
          AB451325.1 - 2 - 3 212 94
          AB451454.1 - 2 - 3 212 94
          AF228636.1 - 2 - 3 224 94
          AY523040.1 - 2 - 3 152 94
          BC066254.1 - 2 - 3 230 6971690 94
          BC066255.1 - 2 - 3 230 6971691 94
          BC066256.1 - 2 - 3 230 6971693 94
          BC066257.1 - 2 - 3 230 6971694 94
          BC070338.1 - 2 - 3 259 30418817 94
          DQ231169.1 - 2 - 3 152 94
          DQ861285.1 - 2 - 3 214 94
          S77834.1 - 2 - 3 259 94
          S77835.1 - 2 - 3 201 94
          S82692.1 - 2 - 3 506 94
          U25676.1 - 2 - 3 268 94
          V00564.1 - 2 - 3 259 94
          X01586.1 - 2 - 3 319 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0400011754.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC04001520.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Immune Tolerance Network Mammalian Gene Collection Recommended Assay Homo sapiens Human Chemokine Panel Human Immune Response Human NFkB Pathway Human Hematopoeisis Human IL-2 Pathway Human Signal Transduction Pathways Human Growth Factors Human Cytokine Network Human IL-2 Gene Expression in Activated and Quiescent T-Cell HUMAN 14-3-3 INDUCED INTRACELLULAR SIGNALING HUMAN CREB PATHWAY HUMAN ENOS SIGNALING HUMAN BREAST CANCER REGULATION BY STATHMIN1 HUMAN P53 MEDIATED APOPTOSIS HUMAN IL-6 PATHWAY HUMAN PATHOGENESIS OF RHEUMATOID ARTHRITIS HUMAN CELLULAR APOPTOSIS PATHWAY Inventoried Human_Immune Human_Nfkb Human_Cytokine_Network Human_IL-2_Gene_Expression_in_Activated_and_Quiescent_T-Cells Human_Growth_Factors Human_Chemokine Human_Hematopoisis Human_Signal_Transduction_Pathways Human_IL-2_Pathway Human_Pathogenesis_of_Rheumatoid_Arthritis Human_IL-6_Pathway Human_14-3-3_Induced_Intracellular_Signaling Human_eNOS_Signaling Human_CREB_Pathway Human_p53_Mediated_Apoptosis Human_Cellular_Apoptosis_Pathway Human_Breast_Cancer_Regulation_by_Stathmin1 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene IL2

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          negative regulation of protein phosphorylation
          adaptive immune response
          negative regulation of B cell apoptotic process
          immune response
          cell adhesion
          positive regulation of cytosolic calcium ion concentration
          protein kinase C-activating G-protein coupled receptor signaling pathway
          cell-cell signaling
          positive regulation of cell proliferation
          natural killer cell activation
          T cell differentiation
          positive regulation of cell growth
          positive regulation of B cell proliferation
          positive regulation of interferon-gamma production
          positive regulation of interleukin-17 production
          positive regulation of tissue remodeling
          positive regulation of activated T cell proliferation
          positive regulation of tyrosine phosphorylation of Stat5 protein
          negative regulation of apoptotic process
          positive regulation of GTPase activity
          response to ethanol
          positive regulation of regulatory T cell differentiation
          negative regulation of heart contraction
          positive regulation of transcription from RNA polymerase II promoter
          regulation of T cell homeostatic proliferation
          positive regulation of isotype switching to IgG isotypes
          negative regulation of lymphocyte proliferation
          negative regulation of inflammatory response
          positive regulation of inflammatory response
          positive regulation of immunoglobulin secretion
          positive regulation of dendritic spine development
          extrinsic apoptotic signaling pathway in absence of ligand
          Ras guanyl-nucleotide exchange factor activity
          cytokine activity
          interleukin-2 receptor binding
          growth factor activity
          kinase activator activity
          carbohydrate binding
          kappa-type opioid receptor binding
          glycosphingolipid binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0400011754.hg.1 ENST00000226730.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011754.hg.1 ENST00000226730.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011754.hg.1 ENST00000226730.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011754.hg.1 ENST00000226730.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011754.hg.1 ENST00000226730.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011754.hg.1 ENST00000226730.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011754.hg.1 ENST00000226730.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011754.hg.1 ENST00000226730.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011754.hg.1 ENST00000226730.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011754.hg.1 ENST00000477645.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011754.hg.1 ENST00000477645.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011754.hg.1 ENST00000477645.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011754.hg.1 ENST00000477645.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011754.hg.1 ENST00000477645.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011754.hg.1 ENST00000477645.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011754.hg.1 NM_000586.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011754.hg.1 NM_000586.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011754.hg.1 NM_000586.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011754.hg.1 NM_000586.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011754.hg.1 NM_000586.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011754.hg.1 NM_000586.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011754.hg.1 NM_000586.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011754.hg.1 NM_000586.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04021599.hg.1 ENST00000226730.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04021599.hg.1 NM_000586.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04021599.hg.1 TC04001520.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0400180911.hg.1 ENST00000226730.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180911.hg.1 ENST00000477645.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180911.hg.1 NM_000586.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400180911.hg.1 ENST00000226730.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180911.hg.1 ENST00000477645.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180911.hg.1 NM_000586.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400180911.hg.1 TC0400011754.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0400011754.hg.1 ENST00000226730.4 Ensembl
          Affymetrix Human Clariom D Assay TC0400011754.hg.1 ENST00000226730.4 Ensembl
          Affymetrix Human Clariom D Assay TC0400011754.hg.1 ENST00000226730.4 Ensembl
          Affymetrix Human Clariom D Assay TC0400011754.hg.1 ENST00000226730.4 Ensembl
          Affymetrix Human Clariom D Assay TC0400011754.hg.1 ENST00000226730.4 Ensembl
          Affymetrix Human Clariom D Assay TC0400011754.hg.1 ENST00000226730.4 Ensembl
          Affymetrix Human Clariom D Assay TC0400011754.hg.1 ENST00000226730.4 Ensembl
          Affymetrix Human Clariom D Assay TC0400011754.hg.1 ENST00000226730.4 Ensembl
          Affymetrix Human Clariom D Assay TC0400011754.hg.1 ENST00000226730.4 Ensembl
          Affymetrix Human Clariom D Assay TC0400011754.hg.1 ENST00000477645.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011754.hg.1 ENST00000477645.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011754.hg.1 ENST00000477645.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011754.hg.1 ENST00000477645.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011754.hg.1 ENST00000477645.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011754.hg.1 ENST00000477645.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011754.hg.1 NM_000586.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011754.hg.1 NM_000586.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011754.hg.1 NM_000586.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011754.hg.1 NM_000586.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011754.hg.1 NM_000586.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011754.hg.1 NM_000586.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011754.hg.1 NM_000586.3 Refseq
          Affymetrix Human Clariom D Assay TC0400011754.hg.1 NM_000586.3 Refseq

          Back To Top
           

          Citations (18)

          Intragraft expression of the IL-10 gene is up-regulated in renal protocol biopsies with early interstitial fibrosis, tubular atrophy, and subclinical rejection.
          Authors
          Hueso M, Navarro E, Moreso F, O'Valle F, Perez-Riba M, Del Moral RG, Grinyo JM, Seron D
          Journal
          Am J Pathol - Page No 1696 (Thu Apr 01 00:00:00 UTC 2010)
          Volume (Year)
          176 (2010)
          Product Usage
          -
          Hypercytokinaemia accompanies HIV-tuberculosis immune reconstitution inflammatory syndrome.
          Authors
          Tadokera R, Meintjes G, Skolimowska KH, Wilkinson KA, Matthews K, Seldon R, Chegou NN, Maartens G, Rangaka MX, Rebe K, Walzl G, Wilkinson RJ
          Journal
          Eur Respir J - Page No (Fri Sep 03 00:00:00 UTC 2010)
          Volume (Year)
           (2010)
          Product Usage
          -
          Altered expression of T cell immunoglobulin-mucin (TIM) molecules in bronchoalveolar lavage CD4+ T cells in sarcoidosis.
          Authors
          Idali F, Wahlstrom J, Dahlberg B, Khademi M, Olsson T, Eklund A, Grunewald J
          Journal
          Respir Res - Page No 42 (Thu Jan 01 00:00:00 UTC 2009)
          Volume (Year)
          10 (2009)
          Product Usage
          -
          Sotrastaurin and cyclosporine drug interaction study in healthy subjects.
          Authors
          Kovarik JM, Stitah S, Slade A, Vitaliti A, Straube F, Grenet O, Winter S, Sfikas N, Seiberling M
          Journal
          Biopharm Drug Dispos - Page No 331 (Thu Jul 01 00:00:00 UTC 2010)
          Volume (Year)
          31 (2010)
          Product Usage
          -
          Inflammatory and microRNA Gene Expression as Prognostic Classifiers of Barrett's Associated Esophageal Adenocarcinoma.
          Authors
          Nguyen GH, Schetter AJ, Chou DB, Bowman ED, Zhao R, Hawkes JE, Mathe E, Kumamoto K, Zhao Y, Budhu A, Hagiwara N, Wang XW, Miyashita M, Casson AG, Harris CC
          Journal
          Clin Cancer Res - Page No (Mon Oct 25 00:00:00 UTC 2010)
          Volume (Year)
           (2010)
          Product Usage
          -
          The effect of the JAK inhibitor CP-690,550 on peripheral immune parameters in stable kidney allograft patients.
          Authors
          van Gurp EA, Schoordijk-Verschoor W, Klepper M, Korevaar SS, Chan G, Weimar W, Baan CC
          Journal
          Transplantation - Page No 79 (Thu Jan 15 00:00:00 UTC 2009)
          Volume (Year)
          87 (2009)
          Product Usage
          -
          Requirement for O-linked N-acetylglucosaminyltransferase in lymphocytes activation.
          Authors
          Golks A, Tran TT, Goetschy JF, Guerini D
          Journal
          Embo J - Page No 4368 (Wed Oct 17 00:00:00 UTC 2007)
          Volume (Year)
          26 (2007)
          Product Usage
          -
          Induction of Foxp3+ regulatory T cells with histone deacetylase inhibitors.
          Authors
          Lucas JL, Mirshahpanah P, Haas-Stapleton E, Asadullah K, Zollner TM, Numerof RP
          Journal
          Cell Immunol - Page No 97 (Thu Jan 01 00:00:00 UTC 2009)
          Volume (Year)
          257 (2009)
          Product Usage
          -
          Differential expression patterns of cytokines in complex regional pain syndrome.
          Authors
          Uceyler N, Eberle T, Rolke R, Birklein F, Sommer C
          Journal
          Pain - Page No 195 (Thu Nov 01 00:00:00 UTC 2007)
          Volume (Year)
          132 (2007)
          Product Usage
          -
          Intragraft FOXP3 mRNA expression reflects antidonor immune reactivity in cardiac allograft patients.
          Authors
          Dijke IE, Velthuis JH, Caliskan K, Korevaar SS, Maat AP, Zondervan PE, Balk AH, Weimar W, Baan CC
          Journal
          Transplantation - Page No 1477 (Fri Jun 15 00:00:00 UTC 2007)
          Volume (Year)
          83 (2007)
          Product Usage
          -
          Soluble Ig-like transcript 3 inhibits tumor allograft rejection in humanized SCID mice and T cell responses in cancer patients.
          Authors
          Suciu-Foca N, Feirt N, Zhang QY, Vlad G, Liu Z, Lin H, Chang CC, Ho EK, Colovai AI, Kaufman H, D'Agati VD, Thaker HM, Remotti H, Galluzzo S, Cinti P, Rabitti C, Allendorf J, Chabot J, Caricato M, Coppola R, Berloco P, Cortesini R
          Journal
          J Immunol - Page No 7432 (Fri Jun 01 00:00:00 UTC 2007)
          Volume (Year)
          178 (2007)
          Product Usage
          -
          Mycolactone suppresses T cell responsiveness by altering both early signaling and posttranslational events.
          Authors
          Boulkroun S, Guenin-Mace L, Thoulouze MI, Monot M, Merckx A, Langsley G, Bismuth G, Di Bartolo V, Demangel C
          Journal
          J Immunol - Page No 1436 (Mon Feb 01 00:00:00 UTC 2010)
          Volume (Year)
          184 (2010)
          Product Usage
          -
          End-stage renal failure and regulatory activities of CD4+CD25bright+FoxP3+ T-cells.
          Authors
          Hendrikx TK, van Gurp EA, Mol WM, Schoordijk W, Sewgobind VD, Ijzermans JN, Weimar W, Baan CC
          Journal
          Nephrol Dial Transplant - Page No 1969 (Mon Jun 01 00:00:00 UTC 2009)
          Volume (Year)
          24 (2009)
          Product Usage
          -
          Essential role of IL-21 in B cell activation, expansion, and plasma cell generation during CD4+ T cell-B cell collaboration.
          Authors
          Kuchen S, Robbins R, Sims GP, Sheng C, Phillips TM, Lipsky PE, Ettinger R
          Journal
          J Immunol - Page No 5886 (Thu Nov 01 00:00:00 UTC 2007)
          Volume (Year)
          179 (2007)
          Product Usage
          -
          Interplay between human adipocytes and T lymphocytes in obesity: CCL20 as an adipochemokine and T lymphocytes as lipogenic modulators.
          Authors
          Duffaut C, Zakaroff-Girard A, Bourlier V, Decaunes P, Maumus M, Chiotasso P, Sengenes C, Lafontan M, Galitzky J, Bouloumie A
          Journal
          Arterioscler Thromb Vasc Biol - Page No 1608 (Thu Oct 01 00:00:00 UTC 2009)
          Volume (Year)
          29 (2009)
          Product Usage
          -
          Characterization of rabbit antithymocyte globulins-induced CD25+ regulatory T cells from cells of patients with end-stage renal disease.
          Authors
          Sewgobind VD, van der Laan LJ, Kho MM, Kraaijeveld R, Korevaar SS, van Dam T, Ijzermans JN, Weimar W, Baan CC
          Journal
          Transplantation - Page No 655 (Sat Mar 27 00:00:00 UTC 2010)
          Volume (Year)
          89 (2010)
          Product Usage
          -
          ITPKC functional polymorphism associated with Kawasaki disease susceptibility and formation of coronary artery aneurysms.
          Authors
          Onouchi Y, Gunji T, Burns JC, Shimizu C, Newburger JW, Yashiro M, Nakamura Y, Yanagawa H, Wakui K, Fukushima Y, Kishi F, Hamamoto K, Terai M, Sato Y, Ouchi K, Saji T, Nariai A, Kaburagi Y, Yoshikawa T, Suzuki K, Tanaka T, Nagai T, Cho H, Fujino A, Sekine A, Nakamichi R, Tsunoda T, Kawasaki T, Nakamura Y, Hata A
          Journal
          Nat Genet - Page No 35 (Tue Jan 01 00:00:00 UTC 2008)
          Volume (Year)
          40 (2008)
          Product Usage
          -
          Local and systemic cytokine and chemokine responses after parenteral influenza vaccination.
          Authors
          Eriksson JC, Cox RJ, Szyszko E, Davidsson A, Brokstad KA
          Journal
          Influenza Other Respi Viruses - Page No 139 (Sun Jul 01 00:00:00 UTC 2007)
          Volume (Year)
          1 (2007)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline