Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00175195_m1
          See other CTSG GE Assays ›
          Gene Symbol:
          CTSG
          Gene Name:
          cathepsin G
          Gene Aliases:
          CATG, CG
          Chromosome Location:
          Chr.14: 24573518 - 24576260 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00175195_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CTSG
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001911.2 NP_001902.1 2 - 3 240 90
          XM_011536499.1 XP_011534801.1 2 - 3 260 90
          GenBank mRNA BC014460.1 - 2 - 3 240 4859911 90
          CR456807.1 - 2 - 3 203 90
          CR541704.1 - 2 - 3 203 90
          M16117.1 - 2 - 3 211 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1400008762.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC14000980.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protease Immune Tolerance Network Mammalian Gene Collection Recommended Assay Homo sapiens Human Alzheimer's Disease Inventoried Human_Alzheimer Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CTSG

          Panther Classification:

          Molecular Function -

          protease serine protease protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          angiotensin maturation
          protein phosphorylation
          proteolysis
          immune response
          protein processing
          extracellular matrix disassembly
          response to lipopolysaccharide
          negative regulation of growth of symbiont in host
          cellular protein metabolic process
          positive regulation of immune response
          defense response to fungus
          neutrophil mediated killing of gram-positive bacterium
          serine-type endopeptidase activity
          protein binding
          heparin binding
          peptidase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR14011250.hg.1 ENST00000216336.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14011250.hg.1 NM_001911.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14011250.hg.1 TC14000980.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1400008762.hg.1 ENST00000216336.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008762.hg.1 ENST00000216336.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008762.hg.1 ENST00000216336.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008762.hg.1 ENST00000216336.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008762.hg.1 ENST00000216336.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008762.hg.1 ENST00000216336.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008762.hg.1 ENST00000216336.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008762.hg.1 ENST00000552252.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008762.hg.1 ENST00000552252.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008762.hg.1 ENST00000552252.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008762.hg.1 ENST00000552252.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008762.hg.1 ENST00000552252.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008762.hg.1 ENST00000552252.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008762.hg.1 ENST00000552252.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008762.hg.1 ENST00000552252.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008762.hg.1 ENST00000552252.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008762.hg.1 NM_001911.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008762.hg.1 NM_001911.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008762.hg.1 NM_001911.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008762.hg.1 NM_001911.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008762.hg.1 NM_001911.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008762.hg.1 NM_001911.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008762.hg.1 NM_001911.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008762.hg.1 NM_001911.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14011251.hg.1 ENST00000216336.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14011251.hg.1 NM_001911.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14011251.hg.1 TC14000980.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR14011249.hg.1 ENST00000216336.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14011249.hg.1 NM_001911.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14011249.hg.1 TC14000980.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1400164559.hg.1 ENST00000216336.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1400164559.hg.1 ENST00000552252.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400164559.hg.1 NM_001911.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400164559.hg.1 ENST00000216336.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1400164559.hg.1 ENST00000552252.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400164559.hg.1 NM_001911.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400164559.hg.1 TC1400008762.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400164560.hg.1 ENST00000216336.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1400164560.hg.1 ENST00000552252.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400164560.hg.1 NM_001911.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400164560.hg.1 ENST00000216336.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1400164560.hg.1 ENST00000552252.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400164560.hg.1 NM_001911.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400164560.hg.1 TC1400008762.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400164557.hg.1 ENST00000216336.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1400164557.hg.1 ENST00000552252.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400164557.hg.1 NM_001911.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400164557.hg.1 ENST00000216336.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1400164557.hg.1 ENST00000552252.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400164557.hg.1 NM_001911.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400164557.hg.1 TC1400008762.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1400008762.hg.1 ENST00000216336.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008762.hg.1 ENST00000216336.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008762.hg.1 ENST00000216336.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008762.hg.1 ENST00000216336.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008762.hg.1 ENST00000216336.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008762.hg.1 ENST00000216336.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008762.hg.1 ENST00000216336.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008762.hg.1 ENST00000552252.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008762.hg.1 ENST00000552252.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008762.hg.1 ENST00000552252.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008762.hg.1 ENST00000552252.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008762.hg.1 ENST00000552252.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008762.hg.1 ENST00000552252.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008762.hg.1 ENST00000552252.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008762.hg.1 ENST00000552252.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008762.hg.1 ENST00000552252.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008762.hg.1 NM_001911.2 Refseq
          Affymetrix Human Clariom D Assay TC1400008762.hg.1 NM_001911.2 Refseq
          Affymetrix Human Clariom D Assay TC1400008762.hg.1 NM_001911.2 Refseq
          Affymetrix Human Clariom D Assay TC1400008762.hg.1 NM_001911.2 Refseq
          Affymetrix Human Clariom D Assay TC1400008762.hg.1 NM_001911.2 Refseq
          Affymetrix Human Clariom D Assay TC1400008762.hg.1 NM_001911.2 Refseq
          Affymetrix Human Clariom D Assay TC1400008762.hg.1 NM_001911.2 Refseq
          Affymetrix Human Clariom D Assay TC1400008762.hg.1 NM_001911.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400164561.hg.1 ENST00000216336.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1400164561.hg.1 ENST00000552252.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400164561.hg.1 NM_001911.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400164561.hg.1 ENST00000216336.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1400164561.hg.1 ENST00000552252.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400164561.hg.1 NM_001911.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400164561.hg.1 TC1400008762.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline