Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00175480_m1
          See other CTLA4 GE Assays ›
          Gene Symbol:
          CTLA4
          Gene Name:
          cytotoxic T-lymphocyte associated protein 4
          Gene Aliases:
          ALPS5, CD, CD152, CELIAC3, CTLA-4, GRD4, GSE, IDDM12
          Chromosome Location:
          Chr.2: 203867788 - 203873960 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00175480_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 190.28
          190.28
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CTLA4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037631.2 NP_001032720.1 1 - 2 262 93
          NM_005214.4 NP_005205.2 1 - 2 262 93
          GenBank mRNA AF414120.1 - 1 - 2 262 93
          AF486806.1 - 1 - 2 107 93
          AK313732.1 - 1 - 2 247 93
          AY209009.1 - 1 - 2 107 93
          AY792514.1 - 1 - 2 107 93
          BC069566.1 - 1 - 2 202 7262243 93
          BC070162.1 - 1 - 2 263 30417685 93
          BC074842.2 - 1 - 2 133 30915552 93
          BC074893.2 - 1 - 2 138 30915247 93
          CD639535.1 - 1 - 2 266 93
          L15006.1 - 1 - 2 107 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200010525.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02001201.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Immune Tolerance Network Mammalian Gene Collection Recommended Assay Homo sapiens Human Immune Response Human Diabetes 1700 Inventoried Human_Immune Human_Diabetes Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CTLA4

          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          adaptive immune response
          immune response
          cellular response to DNA damage stimulus
          negative regulation of B cell proliferation
          T cell costimulation
          positive regulation of apoptotic process
          negative regulation of regulatory T cell differentiation
          B cell receptor signaling pathway
          protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02018466.hg.1 ENST00000302823.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018466.hg.1 ENST00000541886.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018466.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02018466.hg.1 NM_005214.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02018466.hg.1 TC02001201.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02018468.hg.1 ENST00000302823.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018468.hg.1 ENST00000541886.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018468.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02018468.hg.1 NM_005214.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02018468.hg.1 TC02001201.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02018465.hg.1 ENST00000302823.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018465.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02018465.hg.1 NM_005214.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02018465.hg.1 TC02001201.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000295854.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000295854.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000295854.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000295854.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000427473.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000427473.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000427473.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000427473.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000427473.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000427473.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000427473.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000427473.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000472206.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000472206.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000472206.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000472206.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000487393.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000487393.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 ENST00000487393.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 NM_005214.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 NM_005214.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 NM_005214.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 NM_005214.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 NM_005214.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 NM_005214.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 NM_005214.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 NM_005214.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 NM_005214.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 NM_005214.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 NM_005214.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 NM_005214.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010525.hg.1 NM_005214.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02018469.hg.1 ENST00000295854.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018469.hg.1 ENST00000302823.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018469.hg.1 ENST00000472206.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018469.hg.1 ENST00000487393.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018469.hg.1 ENST00000541886.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02018469.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02018469.hg.1 NM_005214.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02018469.hg.1 TC02001201.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0200174587.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0200174587.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200174587.hg.1 NM_005214.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200174587.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0200174587.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200174587.hg.1 NM_005214.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200174587.hg.1 TC0200010525.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200174586.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0200174586.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200174586.hg.1 NM_005214.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200174586.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0200174586.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200174586.hg.1 NM_005214.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200174586.hg.1 TC0200010525.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200174590.hg.1 ENST00000295854.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0200174590.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0200174590.hg.1 ENST00000472206.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200174590.hg.1 ENST00000487393.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200174590.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200174590.hg.1 NM_005214.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200174590.hg.1 ENST00000295854.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0200174590.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0200174590.hg.1 ENST00000472206.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200174590.hg.1 ENST00000487393.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200174590.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200174590.hg.1 NM_005214.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200174590.hg.1 TC0200010525.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000295854.10 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000295854.10 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000295854.10 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000295854.10 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000427473.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000427473.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000427473.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000427473.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000427473.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000427473.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000427473.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000427473.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000472206.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000472206.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000472206.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000472206.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000487393.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000487393.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 ENST00000487393.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 NM_005214.4 Refseq
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 NM_005214.4 Refseq
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 NM_005214.4 Refseq
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 NM_005214.4 Refseq
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 NM_005214.4 Refseq
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 NM_005214.4 Refseq
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 NM_005214.4 Refseq
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 NM_005214.4 Refseq
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 NM_005214.4 Refseq
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 NM_005214.4 Refseq
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 NM_005214.4 Refseq
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 NM_005214.4 Refseq
          Affymetrix Human Clariom D Assay TC0200010525.hg.1 NM_005214.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200174589.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0200174589.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200174589.hg.1 NM_005214.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200174589.hg.1 ENST00000302823.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0200174589.hg.1 NM_001037631.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200174589.hg.1 NM_005214.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200174589.hg.1 TC0200010525.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 167389 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-p58g2:80/100.66.72.198:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0