Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00175732_m1
          See other P2RY2 GE Assays ›
          Gene Symbol:
          P2RY2
          Gene Name:
          purinergic receptor P2Y2
          Gene Aliases:
          HP2U, P2RU1, P2U, P2U1, P2UR, P2Y2, P2Y2R
          Chromosome Location:
          Chr.11: 73200416 - 73246743 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00175732_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: P2RY2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002564.3 NP_002555.3 2 - 3 328 94
          NM_176071.2 NP_788085.2 2 - 3 393 94
          XM_005274019.4 XP_005274076.1 2 - 3 280 94
          XM_005274020.4 XP_005274077.1 2 - 3 1867 94
          XM_005274021.4 XP_005274078.1 2 - 3 1461 94
          XM_011545074.2 XP_011543376.1 2 - 3 215 94
          XM_017017839.1 XP_016873328.1 3 - 4 340 94
          GenBank mRNA BC028135.1 - 2 - 3 393 5215817 94
          U07225.1 - 2 - 3 239 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100008378.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11000772.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the BC028135.1,NM_002564.3,NM_176071.2,U07225.1,XM_005274019.4,XM_005274020.4,XM_005274021.4,XM_011545074.2,XM_017017839.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene P2RY2

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          cellular ion homeostasis
          phospholipase C-activating G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          aging
          positive regulation of neuron projection development
          positive regulation of smooth muscle cell migration
          sensory perception of pain
          negative regulation of ossification
          positive regulation of prostaglandin secretion
          positive regulation of chemokine production
          G-protein coupled purinergic nucleotide receptor signaling pathway
          wound healing
          regulation of vasodilation
          positive regulation of penile erection
          positive regulation of mucus secretion
          response to growth factor
          cellular response to lipopolysaccharide
          cellular response to interleukin-1
          cellular response to purine-containing compound
          receptor activity
          ATP binding
          G-protein coupled purinergic nucleotide receptor activity
          UTP-activated nucleotide receptor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000393596.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000393596.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000393596.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000393596.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000393597.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000393597.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000393597.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000393597.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000565433.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 ENST00000565433.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11009919.hg.1 ENST00000311131.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11009919.hg.1 ENST00000393597.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11009919.hg.1 NM_002564.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11009919.hg.1 NM_176072.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11009919.hg.1 TC11000772.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR11009924.hg.1 ENST00000311131.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11009924.hg.1 ENST00000393596.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11009924.hg.1 ENST00000393597.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11009924.hg.1 NM_002564.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11009924.hg.1 NM_176071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11009924.hg.1 NM_176072.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11009924.hg.1 TC11000772.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1100161922.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100161922.hg.1 ENST00000393596.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100161922.hg.1 ENST00000393597.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100161922.hg.1 NM_002564.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100161922.hg.1 NM_176071.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100161922.hg.1 NM_176072.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100161922.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100161922.hg.1 ENST00000393596.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100161922.hg.1 ENST00000393597.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100161922.hg.1 NM_002564.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100161922.hg.1 NM_176071.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100161922.hg.1 NM_176072.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100161922.hg.1 TC1100008378.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000393596.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000393596.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000393596.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000393596.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000393597.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000393597.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000393597.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000393597.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000565433.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 ENST00000565433.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_002564.3 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176071.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008378.hg.1 NM_176072.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100161917.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100161917.hg.1 ENST00000393597.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100161917.hg.1 NM_002564.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100161917.hg.1 NM_176072.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100161917.hg.1 ENST00000311131.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100161917.hg.1 ENST00000393597.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100161917.hg.1 NM_002564.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100161917.hg.1 NM_176072.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100161917.hg.1 TC1100008378.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline