| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20009104.hg.1 |
ENST00000341420.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20009104.hg.1 |
ENST00000378053.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20009104.hg.1 |
ENST00000541882.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20009104.hg.1 |
NM_013281.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20009104.hg.1 |
NM_198391.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20009104.hg.1 |
TC20000642.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20009103.hg.1 |
ENST00000341420.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20009103.hg.1 |
ENST00000378053.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20009103.hg.1 |
ENST00000541882.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20009103.hg.1 |
NM_013281.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20009103.hg.1 |
NM_198391.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20009103.hg.1 |
TC20000642.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
ENST00000341420.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
ENST00000341420.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
ENST00000341420.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
ENST00000341420.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
ENST00000341420.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
ENST00000341420.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
ENST00000341420.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
ENST00000378053.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
ENST00000378053.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
ENST00000378053.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
ENST00000378053.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
ENST00000378053.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
ENST00000378053.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
ENST00000378053.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
ENST00000462077.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
ENST00000462077.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
ENST00000462077.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
ENST00000462077.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
NM_013281.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
NM_013281.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
NM_013281.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
NM_013281.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
NM_013281.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
NM_013281.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
NM_013281.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
NM_198391.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
NM_198391.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
NM_198391.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
NM_198391.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
NM_198391.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
NM_198391.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
NM_198391.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008445.hg.1 |
NM_198391.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000159389.hg.1 |
ENST00000341420.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000159389.hg.1 |
ENST00000378053.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000159389.hg.1 |
NM_013281.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000159389.hg.1 |
NM_198391.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000159389.hg.1 |
TC2000008445.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000159389.hg.1 |
ENST00000341420.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000159389.hg.1 |
ENST00000378053.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000159389.hg.1 |
NM_013281.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000159389.hg.1 |
NM_198391.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000159390.hg.1 |
ENST00000341420.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000159390.hg.1 |
ENST00000378053.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000159390.hg.1 |
NM_013281.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000159390.hg.1 |
NM_198391.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000159390.hg.1 |
TC2000008445.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000159390.hg.1 |
ENST00000341420.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000159390.hg.1 |
ENST00000378053.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000159390.hg.1 |
NM_013281.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000159390.hg.1 |
NM_198391.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
ENST00000341420.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
ENST00000341420.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
ENST00000341420.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
ENST00000341420.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
ENST00000341420.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
ENST00000341420.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
ENST00000341420.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
ENST00000378053.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
ENST00000378053.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
ENST00000378053.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
ENST00000378053.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
ENST00000378053.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
ENST00000378053.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
ENST00000378053.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
ENST00000462077.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
ENST00000462077.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
ENST00000462077.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
ENST00000462077.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
NM_013281.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
NM_013281.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
NM_013281.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
NM_013281.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
NM_013281.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
NM_013281.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
NM_013281.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
NM_198391.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
NM_198391.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
NM_198391.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
NM_198391.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
NM_198391.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
NM_198391.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
NM_198391.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008445.hg.1 |
NM_198391.2 |
Refseq |