| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000040877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000462259.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000462259.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000462259.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000462259.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000462259.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000462259.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000462259.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000462259.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000462259.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000462259.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000462259.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000462259.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000462259.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000462259.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000463793.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000463793.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000463793.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000463793.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000468077.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000468077.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000468077.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000468077.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000468077.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000471918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000471918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000471918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000481183.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000481183.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000481183.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000481183.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000481183.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000481183.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000481183.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000481183.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000483404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000483404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000483404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000483404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000483404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000483404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000483404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000483404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000483404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000483404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000483404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000483404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000483404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000483404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000483404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000484454.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000484454.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000484454.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000496673.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000496673.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000496673.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000496673.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000496673.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
ENST00000496673.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017788.hg.1 |
NM_005646.3 |
Refseq |