Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00198106_m1
          See other TNFSF13B GE Assays ›
          Gene Symbol:
          TNFSF13B
          Gene Name:
          tumor necrosis factor superfamily member 13b
          Gene Aliases:
          BAFF, BLYS, CD257, DTL, TALL-1, TALL1, THANK, TNFSF20, TNLG7A, ZTNF4
          Chromosome Location:
          Chr.13: 108268240 - 108308484 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00198106_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TNFSF13B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145645.2 NP_001139117.1 4 - 5 956 84
          NM_006573.4 NP_006564.1 5 - 6 1013 84
          GenBank mRNA AF116456.1 - 5 - 6 864 84
          AF132600.1 - 5 - 6 882 84
          AF134715.1 - 5 - 6 890 84
          AF136293.1 - 5 - 6 746 84
          AF186114.1 - 5 - 6 903 84
          AK309629.1 - 6 - 7 783 84
          AY129225.1 - 5 - 6 746 84
          AY302751.1 - 4 - 5 689 84
          AY358881.1 - 5 - 6 915 84
          BC020674.1 - 5 - 6 925 4719315 84
          CR541818.1 - 5 - 6 746 84
          D79690.1 - 1 - 2 83 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1300010000.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC13000383.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Immune Tolerance Network Mammalian Gene Collection Recommended Assay Homo sapiens Human Inflammation Human TNF Superfamily Pathway Inventoried Human_Inflammation Human_TNF_Superfamily_Pathway Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TNFSF13B

          Gene Ontology Categories:

          Function(s) Process(es)

          immune response
          signal transduction
          cell proliferation
          positive regulation of cell proliferation
          positive regulation of B cell proliferation
          tumor necrosis factor-mediated signaling pathway
          receptor binding
          cytokine activity
          tumor necrosis factor receptor binding
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000375887.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000375887.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000375887.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000375887.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000375887.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000375887.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000375887.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000375887.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000375887.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000375887.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000430559.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000430559.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000430559.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000430559.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000430559.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000430559.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000430559.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000430559.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000430559.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000430559.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000479435.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000479435.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000479435.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000479435.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000479435.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000479435.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000479435.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000493765.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000493765.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000493765.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000493765.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000493765.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000542136.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000542136.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 ENST00000542136.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 NM_001145645.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 NM_001145645.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 NM_001145645.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 NM_001145645.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 NM_001145645.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 NM_001145645.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 NM_001145645.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 NM_001145645.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 NM_001145645.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 NM_001145645.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 NM_006573.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 NM_006573.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 NM_006573.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 NM_006573.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 NM_006573.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 NM_006573.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 NM_006573.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 NM_006573.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 NM_006573.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 NM_006573.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300010000.hg.1 NM_006573.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13004134.hg.1 NM_001145645.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13004134.hg.1 NM_006573.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13004134.hg.1 TC13000383.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR13004130.hg.1 ENST00000375887.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13004130.hg.1 ENST00000430559.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13004130.hg.1 NM_001145645.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13004130.hg.1 NM_006573.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13004130.hg.1 TC13000383.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000375887.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000375887.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000375887.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000375887.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000375887.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000375887.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000375887.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000375887.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000375887.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000375887.8 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000430559.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000430559.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000430559.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000430559.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000430559.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000430559.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000430559.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000430559.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000430559.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000430559.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000479435.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000479435.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000479435.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000479435.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000479435.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000479435.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000479435.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000493765.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000493765.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000493765.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000493765.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000493765.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000542136.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000542136.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 ENST00000542136.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 NM_001145645.2 Refseq
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 NM_001145645.2 Refseq
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 NM_001145645.2 Refseq
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 NM_001145645.2 Refseq
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 NM_001145645.2 Refseq
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 NM_001145645.2 Refseq
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 NM_001145645.2 Refseq
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 NM_001145645.2 Refseq
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 NM_001145645.2 Refseq
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 NM_001145645.2 Refseq
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 NM_006573.4 Refseq
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 NM_006573.4 Refseq
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 NM_006573.4 Refseq
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 NM_006573.4 Refseq
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 NM_006573.4 Refseq
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 NM_006573.4 Refseq
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 NM_006573.4 Refseq
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 NM_006573.4 Refseq
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 NM_006573.4 Refseq
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 NM_006573.4 Refseq
          Affymetrix Human Clariom D Assay TC1300010000.hg.1 NM_006573.4 Refseq
          Affymetrix Human Clariom D Assay PSR1300167074.hg.1 ENST00000375887.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1300167074.hg.1 ENST00000430559.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1300167074.hg.1 ENST00000493765.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300167074.hg.1 NM_001145645.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300167074.hg.1 NM_006573.4 Refseq
          Affymetrix Human Clariom D Assay PSR1300167074.hg.1 ENST00000375887.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1300167074.hg.1 ENST00000430559.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1300167074.hg.1 ENST00000493765.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300167074.hg.1 NM_001145645.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300167074.hg.1 NM_006573.4 Refseq
          Affymetrix Human Clariom D Assay PSR1300167074.hg.1 TC1300010000.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300167076.hg.1 ENST00000375887.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1300167076.hg.1 ENST00000430559.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1300167076.hg.1 NM_001145645.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300167076.hg.1 NM_006573.4 Refseq
          Affymetrix Human Clariom D Assay PSR1300167076.hg.1 ENST00000375887.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1300167076.hg.1 ENST00000430559.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1300167076.hg.1 NM_001145645.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300167076.hg.1 NM_006573.4 Refseq
          Affymetrix Human Clariom D Assay PSR1300167076.hg.1 TC1300010000.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300167073.hg.1 ENST00000375887.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1300167073.hg.1 ENST00000430559.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1300167073.hg.1 ENST00000493765.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300167073.hg.1 NM_001145645.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300167073.hg.1 NM_006573.4 Refseq
          Affymetrix Human Clariom D Assay PSR1300167073.hg.1 ENST00000375887.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1300167073.hg.1 ENST00000430559.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1300167073.hg.1 ENST00000493765.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300167073.hg.1 NM_001145645.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300167073.hg.1 NM_006573.4 Refseq
          Affymetrix Human Clariom D Assay PSR1300167073.hg.1 TC1300010000.hg.1 Clariom D TC

          Back To Top
           

          Citations (1)

          Nuclear localization in the biology of the CD40 receptor in normal and neoplastic human B lymphocytes.
          Authors
          Lin-Lee YC, Pham LV, Tamayo AT, Fu L, Zhou HJ, Yoshimura LC, Decker GL, Ford RJ
          Journal
          J Biol Chem - Page No 18878 (Fri Jul 07 00:00:00 UTC 2006)
          Volume (Year)
          281 (2006)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline