| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000424703.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000424703.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000424703.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000424703.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000424703.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000424703.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000424703.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000424703.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000424703.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000424703.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000424703.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000424703.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000424703.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000424703.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000464690.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000464690.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000464690.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000464690.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000464690.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000464690.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000467994.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000467994.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000467994.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000467994.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000468098.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000468098.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000468098.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000468098.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000468098.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000469005.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000469005.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000469005.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000469005.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000469005.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000469005.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000469005.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000472536.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000472536.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000472536.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000480823.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000480823.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000480823.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000480823.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000485968.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000485968.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000485968.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000488318.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000488318.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
ENST00000488318.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012145.hg.1 |
NR_029584.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03030513.hg.1 |
ENST00000295633.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03030513.hg.1 |
ENST00000539471.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03030513.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03030513.hg.1 |
TC03001691.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000424703.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000424703.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000424703.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000424703.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000424703.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000424703.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000424703.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000424703.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000424703.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000424703.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000424703.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000424703.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000424703.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000424703.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000464690.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000464690.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000464690.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000464690.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000464690.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000464690.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000467994.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000467994.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000467994.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000467994.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000468098.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000468098.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000468098.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000468098.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000468098.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000469005.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000469005.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000469005.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000469005.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000469005.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000469005.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000469005.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000472536.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000472536.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000472536.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000480823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000480823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000480823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000480823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000485968.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000485968.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000485968.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000488318.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000488318.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
ENST00000488318.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012145.hg.1 |
NR_029584.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300190397.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300190397.hg.1 |
ENST00000464690.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300190397.hg.1 |
ENST00000469005.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300190397.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300190397.hg.1 |
ENST00000295633.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300190397.hg.1 |
ENST00000464690.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300190397.hg.1 |
ENST00000469005.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300190397.hg.1 |
NM_007085.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300190397.hg.1 |
TC0300012145.hg.1 |
Clariom D TC |