| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006861.hg.1 |
ENSTR0000326153.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006861.hg.1 |
ENSTR0000326153.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006861.hg.1 |
ENSTR0000326153.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006861.hg.1 |
ENSTR0000326153.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006861.hg.1 |
ENSTR0000326153.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006861.hg.1 |
ENSTR0000326153.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006861.hg.1 |
ENSTR0000326153.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006861.hg.1 |
ENSTR0000326153.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006861.hg.1 |
ENSTR0000326153.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006861.hg.1 |
ENSTR0000326153.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006861.hg.1 |
ENSTR0000326153.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006861.hg.1 |
ENSTR0000326153.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006861.hg.1 |
ENSTR0000326153.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006861.hg.1 |
ENSTR0000485332.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006861.hg.1 |
ENSTR0000485332.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006861.hg.1 |
ENSTR0000485332.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006861.hg.1 |
ENSTR0000485332.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006861.hg.1 |
ENSTR0000485332.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006861.hg.1 |
ENSTR0000485332.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006861.hg.1 |
ENSTR0000485332.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
ENST00000326153.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
ENST00000326153.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
ENST00000326153.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
ENST00000326153.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
ENST00000326153.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
ENST00000326153.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
ENST00000326153.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
ENST00000326153.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
ENST00000326153.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
ENST00000326153.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
ENST00000326153.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
ENST00000326153.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
ENST00000326153.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
ENST00000485332.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
ENST00000485332.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
ENST00000485332.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
ENST00000485332.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
ENST00000485332.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
ENST00000485332.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
ENST00000485332.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
NM_012227.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
NM_012227.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
NM_012227.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
NM_012227.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
NM_012227.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
NM_012227.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
NM_012227.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
NM_012227.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
NM_012227.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
NM_012227.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
NM_012227.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
NM_012227.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008886.hg.1 |
NM_012227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
ENST00000326153.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
ENST00000326153.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
ENST00000326153.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
ENST00000326153.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
ENST00000326153.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
ENST00000326153.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
ENST00000326153.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
ENST00000326153.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
ENST00000326153.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
ENST00000326153.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
ENST00000326153.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
ENST00000326153.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
ENST00000326153.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
ENST00000485332.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
ENST00000485332.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
ENST00000485332.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
ENST00000485332.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
ENST00000485332.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
ENST00000485332.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
ENST00000485332.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
NM_012227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
NM_012227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
NM_012227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
NM_012227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
NM_012227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
NM_012227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
NM_012227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
NM_012227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
NM_012227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
NM_012227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
NM_012227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
NM_012227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008886.hg.1 |
NM_012227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0Y00006861.hg.1 |
ENSTR0000326153.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00006861.hg.1 |
ENSTR0000326153.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00006861.hg.1 |
ENSTR0000326153.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00006861.hg.1 |
ENSTR0000326153.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00006861.hg.1 |
ENSTR0000326153.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00006861.hg.1 |
ENSTR0000326153.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00006861.hg.1 |
ENSTR0000326153.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00006861.hg.1 |
ENSTR0000326153.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00006861.hg.1 |
ENSTR0000326153.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00006861.hg.1 |
ENSTR0000326153.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00006861.hg.1 |
ENSTR0000326153.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00006861.hg.1 |
ENSTR0000326153.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00006861.hg.1 |
ENSTR0000326153.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00006861.hg.1 |
ENSTR0000485332.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00006861.hg.1 |
ENSTR0000485332.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00006861.hg.1 |
ENSTR0000485332.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00006861.hg.1 |
ENSTR0000485332.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00006861.hg.1 |
ENSTR0000485332.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00006861.hg.1 |
ENSTR0000485332.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00006861.hg.1 |
ENSTR0000485332.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00161059.hg.1 |
ENST00000326153.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00161059.hg.1 |
ENST00000485332.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00161059.hg.1 |
NM_012227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00161059.hg.1 |
ENST00000326153.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00161059.hg.1 |
ENST00000485332.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00161059.hg.1 |
NM_012227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00161059.hg.1 |
TC0X00008886.hg.1 |
Clariom D TC |