Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00219866_m1
          See other CYP26B1 GE Assays ›
          Gene Symbol:
          CYP26B1
          Gene Name:
          cytochrome P450 family 26 subfamily B member 1
          Gene Aliases:
          CYP26A2, P450RAI-2, P450RAI2, RHFCA
          Chromosome Location:
          Chr.2: 72129238 - 72147862 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00219866_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CYP26B1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019885.3 NP_063938.1 1 - 2 230 89
          GenBank mRNA AF252297.1 - 1 - 2 202 89
          AK313433.1 - 1 - 2 205 89
          BC069443.1 - 1 - 2 202 7262360 89
          BC109205.2 - 1 - 2 202 40006245 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200013042.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02001976.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection P450 Homo sapiens Human CYP450 and other Oxygenases Inventoried Human CYP450 + oxygenases Human_CYP450_and_other_Oxygenases Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CYP26B1

          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          cell fate determination
          establishment of T cell polarity
          vitamin metabolic process
          xenobiotic metabolic process
          inflammatory response
          male meiosis
          spermatogenesis
          proximal/distal pattern formation
          positive regulation of gene expression
          embryonic limb morphogenesis
          retinoic acid catabolic process
          tongue morphogenesis
          regulation of T cell differentiation
          retinoic acid receptor signaling pathway
          negative regulation of retinoic acid receptor signaling pathway
          oxidation-reduction process
          bone morphogenesis
          establishment of skin barrier
          cornification
          cellular response to retinoic acid
          positive regulation of tongue muscle cell differentiation
          retinoic acid binding
          iron ion binding
          retinoic acid 4-hydroxylase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, NAD(P)H as one donor, and incorporation of one atom of oxygen
          heme binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02031467.hg.1 ENST00000001146.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02031467.hg.1 NM_019885.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02031467.hg.1 TC02001976.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000412253.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000412253.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000412253.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000412253.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000412253.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000412253.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000412253.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000412253.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000412253.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000461519.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000461519.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000461519.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000474509.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000474509.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000474509.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000474509.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000474509.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000546307.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000546307.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000546307.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000546307.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000546307.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000546307.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000546307.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000546307.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000546307.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000546307.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 ENST00000546307.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 NM_001277742.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 NM_001277742.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 NM_001277742.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 NM_001277742.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 NM_001277742.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 NM_001277742.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 NM_001277742.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 NM_001277742.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 NM_001277742.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 NM_001277742.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 NM_001277742.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 NM_019885.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 NM_019885.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 NM_019885.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 NM_019885.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 NM_019885.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 NM_019885.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 NM_019885.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 NM_019885.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 NM_019885.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 NM_019885.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 NM_019885.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013042.hg.1 NM_019885.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02031469.hg.1 ENST00000001146.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02031469.hg.1 ENST00000546307.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02031469.hg.1 NM_019885.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02031469.hg.1 TC02001976.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0200195199.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200195199.hg.1 ENST00000474509.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200195199.hg.1 ENST00000546307.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200195199.hg.1 NM_001277742.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200195199.hg.1 NM_019885.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200195199.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200195199.hg.1 ENST00000474509.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200195199.hg.1 ENST00000546307.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200195199.hg.1 NM_001277742.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200195199.hg.1 NM_019885.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200195199.hg.1 TC0200013042.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000412253.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000412253.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000412253.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000412253.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000412253.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000412253.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000412253.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000412253.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000412253.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000461519.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000461519.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000461519.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000474509.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000474509.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000474509.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000474509.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000474509.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000546307.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000546307.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000546307.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000546307.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000546307.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000546307.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000546307.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000546307.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000546307.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000546307.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 ENST00000546307.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 NM_001277742.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 NM_001277742.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 NM_001277742.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 NM_001277742.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 NM_001277742.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 NM_001277742.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 NM_001277742.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 NM_001277742.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 NM_001277742.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 NM_001277742.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 NM_001277742.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 NM_019885.3 Refseq
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 NM_019885.3 Refseq
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 NM_019885.3 Refseq
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 NM_019885.3 Refseq
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 NM_019885.3 Refseq
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 NM_019885.3 Refseq
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 NM_019885.3 Refseq
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 NM_019885.3 Refseq
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 NM_019885.3 Refseq
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 NM_019885.3 Refseq
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 NM_019885.3 Refseq
          Affymetrix Human Clariom D Assay TC0200013042.hg.1 NM_019885.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200195196.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200195196.hg.1 ENST00000461519.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200195196.hg.1 NM_019885.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200195196.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200195196.hg.1 ENST00000461519.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200195196.hg.1 NM_019885.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200195196.hg.1 TC0200013042.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200195198.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200195198.hg.1 ENST00000474509.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200195198.hg.1 ENST00000546307.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200195198.hg.1 NM_001277742.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200195198.hg.1 NM_019885.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200195198.hg.1 ENST00000001146.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200195198.hg.1 ENST00000474509.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200195198.hg.1 ENST00000546307.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200195198.hg.1 NM_001277742.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200195198.hg.1 NM_019885.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200195198.hg.1 TC0200013042.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline