Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00226277_m1
          See other RHBDF2 GE Assays ›
          Gene Symbol:
          RHBDF2
          Gene Name:
          rhomboid 5 homolog 2
          Gene Aliases:
          RHBDL5, RHBDL6, TOC, TOCG, iRhom2
          Chromosome Location:
          Chr.17: 76470893 - 76501427 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00226277_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RHBDF2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001005498.4 NP_001005498.2 12 - 13 1757 75
          NM_001376228.1 NP_001363157.1 12 - 13 2174 75
          NM_001376229.1 NP_001363158.1 12 - 13 2397 75
          NM_001376230.1 NP_001363159.1 12 - 13 1969 75
          NM_024599.5 NP_078875.4 12 - 13 1848 75
          NR_164785.1 12 - 13 2186 75
          NR_164786.1 13 - 14 1928 75
          NR_164787.1 12 - 13 2244 75
          XM_005257669.4 XP_005257726.1 12 - 13 2261 75
          XM_011525249.3 XP_011523551.1 12 - 13 1779 75
          XM_011525250.3 XP_011523552.1 13 - 14 1862 75
          XM_011525251.3 XP_011523553.1 12 - 13 1703 75
          XM_047436743.1 XP_047292699.1 12 - 13 2321 75
          XM_047436744.1 XP_047292700.1 12 - 13 1790 75
          XM_047436745.1 XP_047292701.1 13 - 14 1949 75
          XM_047436746.1 XP_047292702.1 13 - 14 2362 75
          XM_047436747.1 XP_047292703.1 12 - 13 2203 75
          XM_047436748.1 XP_047292704.1 13 - 14 1967 75
          XM_047436749.1 XP_047292705.1 12 - 13 1866 75
          XM_047436750.1 XP_047292706.1 13 - 14 2422 75
          XM_047436751.1 XP_047292707.1 13 - 14 2275 75
          XM_047436752.1 XP_047292708.1 12 - 13 2116 75
          XM_047436753.1 XP_047292709.1 13 - 14 1880 75
          XM_047436755.1 XP_047292711.1 12 - 13 2234 75
          XM_047436756.1 XP_047292712.1 13 - 14 2335 75
          GenBank mRNA AK025994.1 - 7 - 8 872 75
          AK074034.1 - 10 - 11 2153 75
          AK225684.1 - 7 - 8 825 75
          AK292135.1 - 12 - 13 1841 75
          AK292166.1 - 11 - 12 1641 75
          AL833680.1 - 6 - 7 741 75
          AY500247.1 - 11 - 12 1537 75
          BC016034.1 - 12 - 13 1828 75
          BC035829.1 - 7 - 8 934 75
          BE076010.1 - 3 - 4 201 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700011802.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17001899.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RHBDF2

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of inflammatory response to antigenic stimulus
          protein transport
          regulation of epidermal growth factor receptor signaling pathway
          regulation of protein secretion
          negative regulation of protein secretion
          protein localization to plasma membrane
          serine-type endopeptidase activity
          protein binding
          growth factor binding
          protein transporter activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR17025782.hg.1 ENST00000313080.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17025782.hg.1 ENST00000389760.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17025782.hg.1 ENST00000389762.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17025782.hg.1 NM_001005498.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17025782.hg.1 NM_024599.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17025782.hg.1 TC17001899.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1700196398.hg.1 ENST00000313080.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1700196398.hg.1 ENST00000590168.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700196398.hg.1 ENST00000591885.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700196398.hg.1 NM_001005498.3 Refseq
          Affymetrix Human Clariom D Assay PSR1700196398.hg.1 NM_024599.5 Refseq
          Affymetrix Human Clariom D Assay PSR1700196398.hg.1 ENST00000313080.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1700196398.hg.1 ENST00000590168.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700196398.hg.1 ENST00000591885.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700196398.hg.1 NM_001005498.3 Refseq
          Affymetrix Human Clariom D Assay PSR1700196398.hg.1 NM_024599.5 Refseq
          Affymetrix Human Clariom D Assay PSR1700196398.hg.1 TC1700011802.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline