Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00232488_m1
          See other LMO4 GE Assays ›
          Gene Symbol:
          LMO4
          Gene Name:
          LIM domain only 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 87328468 - 87348923 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00232488_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LMO4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_006769.3 NP_006760.1 1 - 2 775 61
          GenBank mRNA AK292578.1 - 1 - 2 377 61
          BC003600.1 - 1 - 2 138 3355972 61
          BC017673.1 - 1 - 2 263 3893112 61
          BU580860.1 - 1 - 2 225 61
          U24576.1 - 1 - 2 775 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100008938.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01000830.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK292578.1,BC003600.1,BC017673.1,NM_006769.3,U24576.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LMO4

          Gene Ontology Categories:

          Function(s) Process(es)

          neural tube closure
          ventricular septum development
          transcription from RNA polymerase II promoter
          ventral spinal cord interneuron differentiation
          spinal cord motor neuron differentiation
          spinal cord association neuron differentiation
          regulation of cell migration
          negative regulation of protein complex assembly
          positive regulation of kinase activity
          regulation of cell fate specification
          positive regulation of transcription from RNA polymerase II promoter
          thymus development
          regulation of cell activation
          enhancer sequence-specific DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          transcription factor binding
          zinc ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 ENST00000370542.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 ENST00000370542.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 ENST00000370542.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 ENST00000370542.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 ENST00000370542.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 ENST00000370542.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 ENST00000370542.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 ENST00000370542.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 ENST00000370544.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 ENST00000370544.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 ENST00000370544.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 ENST00000370544.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 ENST00000370544.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 ENST00000370544.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 ENST00000370544.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 ENST00000370544.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 ENST00000370544.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 ENST00000489303.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 ENST00000489303.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 ENST00000489303.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 ENST00000489303.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 ENST00000489303.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 ENST00000489303.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 ENST00000495705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 ENST00000495705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 ENST00000495705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 ENST00000495705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 NM_006769.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 NM_006769.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 NM_006769.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 NM_006769.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 NM_006769.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 NM_006769.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 NM_006769.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 NM_006769.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008938.hg.1 NM_006769.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01013205.hg.1 ENST00000370544.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01013205.hg.1 NM_006769.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01013205.hg.1 TC01000830.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01013213.hg.1 ENST00000370542.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01013213.hg.1 ENST00000370544.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01013213.hg.1 ENST00000489303.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01013213.hg.1 NM_006769.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01013213.hg.1 TC01000830.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01013204.hg.1 ENST00000370544.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01013204.hg.1 NM_006769.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01013204.hg.1 TC01000830.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01013207.hg.1 ENST00000370544.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01013207.hg.1 NM_006769.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01013207.hg.1 TC01000830.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100163799.hg.1 ENST00000370544.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100163799.hg.1 NM_006769.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100163799.hg.1 ENST00000370544.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100163799.hg.1 NM_006769.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100163799.hg.1 TC0100008938.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 ENST00000370542.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 ENST00000370542.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 ENST00000370542.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 ENST00000370542.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 ENST00000370542.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 ENST00000370542.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 ENST00000370542.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 ENST00000370542.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 ENST00000370544.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 ENST00000370544.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 ENST00000370544.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 ENST00000370544.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 ENST00000370544.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 ENST00000370544.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 ENST00000370544.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 ENST00000370544.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 ENST00000370544.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 ENST00000489303.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 ENST00000489303.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 ENST00000489303.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 ENST00000489303.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 ENST00000489303.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 ENST00000489303.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 ENST00000495705.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 ENST00000495705.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 ENST00000495705.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 ENST00000495705.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 NM_006769.3 Refseq
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 NM_006769.3 Refseq
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 NM_006769.3 Refseq
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 NM_006769.3 Refseq
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 NM_006769.3 Refseq
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 NM_006769.3 Refseq
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 NM_006769.3 Refseq
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 NM_006769.3 Refseq
          Affymetrix Human Clariom D Assay TC0100008938.hg.1 NM_006769.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100163812.hg.1 ENST00000370542.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100163812.hg.1 ENST00000370544.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100163812.hg.1 ENST00000489303.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100163812.hg.1 ENST00000495705.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100163812.hg.1 NM_006769.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100163812.hg.1 ENST00000370542.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100163812.hg.1 ENST00000370544.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100163812.hg.1 ENST00000489303.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100163812.hg.1 ENST00000495705.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100163812.hg.1 NM_006769.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100163812.hg.1 TC0100008938.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100163800.hg.1 ENST00000370544.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100163800.hg.1 NM_006769.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100163800.hg.1 ENST00000370544.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100163800.hg.1 NM_006769.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100163800.hg.1 TC0100008938.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100163802.hg.1 ENST00000370544.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100163802.hg.1 ENST00000495705.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100163802.hg.1 NM_006769.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100163802.hg.1 ENST00000370544.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100163802.hg.1 ENST00000495705.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100163802.hg.1 NM_006769.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100163802.hg.1 TC0100008938.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline