Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00232764_m1
          See other FOXA2 GE Assays ›
          Gene Symbol:
          FOXA2
          Gene Name:
          forkhead box A2
          Gene Aliases:
          HNF-3-beta, HNF3B, TCF3B
          Chromosome Location:
          Chr.20: 22580998 - 22585490 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00232764_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FOXA2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021784.5 NP_068556.2 1 - 2 387 66
          NM_153675.3 NP_710141.1 2 - 3 292 66
          XM_047440133.1 XP_047296089.1 2 - 3 255 66
          GenBank mRNA AB028021.1 - 2 - 3 271 66
          BC006545.2 - 1 - 2 71 66
          BC011780.2 - 1 - 2 117 66
          BC019288.1 - 1 - 2 71 66
          BM273279.1 - 1 - 2 285 66
          BQ643619.1 - 266 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000008636.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC20000698.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Human Stem Cell Pluripotency Human Hedgehog Pathway Human Signal Transduction Pathways Human Transcription Factors (non-Hox) Human Maturity-Onset Diabetes of Young (MODY) Inventoried Human_Maturity-Onset_Diabetes_of_Young_(MODY) Human_Signal_Transduction_Pathways Human_Stem_Cell Human_Hedgehog Human_Transcription_Factors_(non-Hox) Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene FOXA2

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          cell fate specification
          chromatin organization
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          adult locomotory behavior
          anatomical structure morphogenesis
          negative regulation of epithelial to mesenchymal transition
          cell differentiation
          regulation of blood coagulation
          endocrine pancreas development
          positive regulation of embryonic development
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          regulation of insulin secretion involved in cellular response to glucose stimulus
          response to interleukin-6
          dopaminergic neuron differentiation
          primitive streak formation
          positive regulation of cell-cell adhesion mediated by cadherin
          positive regulation of gastrulation
          transcription regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          nucleic acid binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription corepressor activity
          protein binding
          protein domain specific binding
          sequence-specific DNA binding
          RNA polymerase II sequence-specific DNA binding transcription factor binding
          sequence-specific double-stranded DNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC2000008636.hg.1 ENST00000377115.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008636.hg.1 ENST00000377115.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008636.hg.1 ENST00000377115.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008636.hg.1 ENST00000377115.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008636.hg.1 ENST00000377115.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008636.hg.1 ENST00000419308.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008636.hg.1 ENST00000419308.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008636.hg.1 ENST00000419308.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008636.hg.1 ENST00000419308.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008636.hg.1 ENST00000419308.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008636.hg.1 NM_021784.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008636.hg.1 NM_021784.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008636.hg.1 NM_021784.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008636.hg.1 NM_021784.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008636.hg.1 NM_021784.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008636.hg.1 NM_153675.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008636.hg.1 NM_153675.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008636.hg.1 NM_153675.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008636.hg.1 NM_153675.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008636.hg.1 NM_153675.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline