Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00236937_m1
          See other CXCL1 GE Assays ›
          Gene Symbol:
          CXCL1
          Gene Name:
          C-X-C motif chemokine ligand 1
          Gene Aliases:
          FSP, GRO1, GROa, MGSA, MGSA-a, NAP-3, SCYB1
          Chromosome Location:
          Chr.4: 73869392 - 73871302 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00236937_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CXCL1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001511.3 NP_001502.1 3 - 4 386 70
          NR_046035.1 3 - 4 409 70
          GenBank mRNA BC011976.1 - 3 - 4 374 3856841 70
          BG121608.1 - 3 - 4 393 70
          BM554448.1 - 3 - 4 399 70
          CB112830.1 - 3 - 4 387 70
          J03561.1 - 3 - 4 360 70
          X12510.1 - 3 - 4 324 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0400007840.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC04000411.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Human Chemokine Panel Human NFkB Pathway Human Growth Factors Inventoried Human_Nfkb Human_Growth_Factors Human_Chemokine Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CXCL1

          Panther Classification:

          Molecular Function -

          chemokine

          Gene Ontology Categories:

          Function(s) Process(es)

          chemotaxis
          inflammatory response
          immune response
          signal transduction
          G-protein coupled receptor signaling pathway
          nervous system development
          cell proliferation
          negative regulation of cell proliferation
          actin cytoskeleton organization
          response to lipopolysaccharide
          intracellular signal transduction
          positive regulation of catalytic activity
          cell chemotaxis
          chemokine-mediated signaling pathway
          positive regulation of neutrophil chemotaxis
          receptor binding
          chemokine activity
          enzyme activator activity
          growth factor activity
          CXCR chemokine receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0400007840.hg.1 ENST00000395761.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007840.hg.1 ENST00000395761.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007840.hg.1 ENST00000395761.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007840.hg.1 ENST00000395761.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007840.hg.1 ENST00000395761.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007840.hg.1 ENST00000509101.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007840.hg.1 ENST00000509101.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007840.hg.1 ENST00000509101.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007840.hg.1 ENST00000509101.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007840.hg.1 ENST00000509101.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007840.hg.1 NM_001511.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007840.hg.1 NM_001511.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007840.hg.1 NM_001511.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007840.hg.1 NM_001511.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007840.hg.1 NM_001511.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007840.hg.1 NM_001511.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007840.hg.1 NR_046035.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007840.hg.1 NR_046035.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007840.hg.1 NR_046035.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007840.hg.1 NR_046035.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007840.hg.1 NR_046035.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007840.hg.1 NR_046035.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04005668.hg.1 NM_001511.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04005668.hg.1 TC04000411.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04005666.hg.1 ENST00000395761.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04005666.hg.1 NM_001511.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04005666.hg.1 TC04000411.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0400153731.hg.1 ENST00000395761.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0400153731.hg.1 NM_001511.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400153731.hg.1 NR_046035.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400153731.hg.1 ENST00000395761.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0400153731.hg.1 NM_001511.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400153731.hg.1 NR_046035.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400153731.hg.1 TC0400007840.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400153733.hg.1 NM_001511.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400153733.hg.1 NR_046035.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400153733.hg.1 NM_001511.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400153733.hg.1 NR_046035.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400153733.hg.1 TC0400007840.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0400007840.hg.1 ENST00000395761.3 Ensembl
          Affymetrix Human Clariom D Assay TC0400007840.hg.1 ENST00000395761.3 Ensembl
          Affymetrix Human Clariom D Assay TC0400007840.hg.1 ENST00000395761.3 Ensembl
          Affymetrix Human Clariom D Assay TC0400007840.hg.1 ENST00000395761.3 Ensembl
          Affymetrix Human Clariom D Assay TC0400007840.hg.1 ENST00000395761.3 Ensembl
          Affymetrix Human Clariom D Assay TC0400007840.hg.1 ENST00000509101.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007840.hg.1 ENST00000509101.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007840.hg.1 ENST00000509101.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007840.hg.1 ENST00000509101.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007840.hg.1 ENST00000509101.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007840.hg.1 NM_001511.3 Refseq
          Affymetrix Human Clariom D Assay TC0400007840.hg.1 NM_001511.3 Refseq
          Affymetrix Human Clariom D Assay TC0400007840.hg.1 NM_001511.3 Refseq
          Affymetrix Human Clariom D Assay TC0400007840.hg.1 NM_001511.3 Refseq
          Affymetrix Human Clariom D Assay TC0400007840.hg.1 NM_001511.3 Refseq
          Affymetrix Human Clariom D Assay TC0400007840.hg.1 NM_001511.3 Refseq
          Affymetrix Human Clariom D Assay TC0400007840.hg.1 NR_046035.1 Refseq
          Affymetrix Human Clariom D Assay TC0400007840.hg.1 NR_046035.1 Refseq
          Affymetrix Human Clariom D Assay TC0400007840.hg.1 NR_046035.1 Refseq
          Affymetrix Human Clariom D Assay TC0400007840.hg.1 NR_046035.1 Refseq
          Affymetrix Human Clariom D Assay TC0400007840.hg.1 NR_046035.1 Refseq
          Affymetrix Human Clariom D Assay TC0400007840.hg.1 NR_046035.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-7fb9f846c9-xq5gc:80/100.66.76.64:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0