Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00253876_m1
          See other NR1D1 GE Assays ›
          Gene Symbol:
          NR1D1
          Gene Name:
          nuclear receptor subfamily 1 group D member 1
          Gene Aliases:
          EAR1, REVERBA, REVERBalpha, THRA1, THRAL, ear-1, hRev
          Chromosome Location:
          Chr.17: 40092793 - 40100589 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00253876_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NR1D1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021724.5 NP_068370.1 1 - 2 529 60
          GenBank mRNA AK315270.1 - 1 - 2 521 60
          BC047875.1 - 1 - 2 489 60
          BC056148.1 - 1 - 2 556 60
          HQ692861.1 - 1 - 2 34 60
          M24898.1 - 1 - 2 661 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700010604.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17001459.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Human Nuclear Receptors 1700 Inventoried Human_Nuclear_Receptors Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene NR1D1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          cellular glucose homeostasis
          glycogen biosynthetic process
          regulation of transcription, DNA-templated
          circadian rhythm
          hormone-mediated signaling pathway
          proteasomal protein catabolic process
          regulation of lipid metabolic process
          cell differentiation
          intracellular steroid hormone receptor signaling pathway
          intracellular receptor signaling pathway
          protein destabilization
          circadian regulation of gene expression
          negative regulation of toll-like receptor 4 signaling pathway
          cholesterol homeostasis
          regulation of circadian sleep/wake cycle
          regulation of circadian rhythm
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          response to leptin
          regulation of fat cell differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          rhythmic process
          negative regulation of inflammatory response
          circadian temperature homeostasis
          regulation of insulin secretion involved in cellular response to glucose stimulus
          regulation of type B pancreatic cell proliferation
          negative regulation of astrocyte activation
          positive regulation of bile acid biosynthetic process
          cellular response to lipopolysaccharide
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          negative regulation of cold-induced thermogenesis
          negative regulation of neuroinflammatory response
          negative regulation of microglial cell activation
          transcription regulatory region sequence-specific DNA binding
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcription corepressor binding
          transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          protein binding
          zinc ion binding
          heme binding
          sequence-specific DNA binding
          metal ion binding
          E-box binding
          sequence-specific double-stranded DNA binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR17019331.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17019331.hg.1 NM_021724.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17019331.hg.1 TC17001459.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR17019333.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17019333.hg.1 NM_021724.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17019333.hg.1 TC17001459.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1700010604.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010604.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010604.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010604.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010604.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010604.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010604.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010604.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010604.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010604.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010604.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010604.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010604.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010604.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010604.hg.1 NM_021724.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010604.hg.1 NM_021724.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010604.hg.1 NM_021724.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010604.hg.1 NM_021724.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010604.hg.1 NM_021724.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010604.hg.1 NM_021724.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010604.hg.1 NM_021724.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010604.hg.1 NM_021724.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010604.hg.1 NM_021724.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010604.hg.1 NM_021724.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010604.hg.1 NM_021724.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010604.hg.1 NM_021724.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010604.hg.1 NM_021724.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010604.hg.1 NM_021724.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17019334.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17019334.hg.1 NM_021724.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17019334.hg.1 TC17001459.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR17019332.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17019332.hg.1 NM_021724.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17019332.hg.1 TC17001459.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1700183646.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700183646.hg.1 NM_021724.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700183646.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700183646.hg.1 NM_021724.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700183646.hg.1 TC1700010604.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700183641.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700183641.hg.1 NM_021724.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700183641.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700183641.hg.1 NM_021724.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700183641.hg.1 TC1700010604.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700183647.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700183647.hg.1 NM_021724.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700183647.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700183647.hg.1 NM_021724.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700183647.hg.1 TC1700010604.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700183643.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700183643.hg.1 NM_021724.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700183643.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700183643.hg.1 NM_021724.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700183643.hg.1 TC1700010604.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700183644.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700183644.hg.1 NM_021724.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700183644.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700183644.hg.1 NM_021724.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700183644.hg.1 TC1700010604.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700183645.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700183645.hg.1 NM_021724.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700183645.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700183645.hg.1 NM_021724.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700183645.hg.1 TC1700010604.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1700010604.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700010604.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700010604.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700010604.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700010604.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700010604.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700010604.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700010604.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700010604.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700010604.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700010604.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700010604.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700010604.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700010604.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700010604.hg.1 NM_021724.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010604.hg.1 NM_021724.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010604.hg.1 NM_021724.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010604.hg.1 NM_021724.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010604.hg.1 NM_021724.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010604.hg.1 NM_021724.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010604.hg.1 NM_021724.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010604.hg.1 NM_021724.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010604.hg.1 NM_021724.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010604.hg.1 NM_021724.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010604.hg.1 NM_021724.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010604.hg.1 NM_021724.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010604.hg.1 NM_021724.4 Refseq
          Affymetrix Human Clariom D Assay TC1700010604.hg.1 NM_021724.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700183640.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700183640.hg.1 NM_021724.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700183640.hg.1 ENST00000246672.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700183640.hg.1 NM_021724.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700183640.hg.1 TC1700010604.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 164860 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline