| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
8100342 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8100344 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8100345 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8100343 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8100341 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8100338 |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
219987_PM_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
ENST00000440542.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
ENST00000440542.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
ENST00000440542.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
ENST00000440542.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
ENST00000440542.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
ENST00000440542.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
ENST00000443173.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
ENST00000443173.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
ENST00000443173.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
ENST00000443173.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
ENST00000443173.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
ENST00000443173.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
ENST00000443173.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
ENST00000443173.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
ENST00000454756.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
ENST00000454756.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
ENST00000454756.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
ENST00000454756.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
NM_001242690.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
NM_001242690.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
NM_001242690.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
NM_001242690.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
NM_001242690.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
NM_001242690.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
NM_001242690.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
NM_024534.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
NM_024534.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
NM_024534.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
NM_024534.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
NM_024534.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
NM_024534.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
NM_024534.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010694.hg.1 |
NM_024534.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04016404.hg.1 |
ENST00000440542.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04016404.hg.1 |
ENST00000443173.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04016404.hg.1 |
ENST00000540758.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04016404.hg.1 |
NM_001242690.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04016404.hg.1 |
NM_024534.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04016404.hg.1 |
TC04001196.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
ENST00000440542.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
ENST00000440542.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
ENST00000440542.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
ENST00000440542.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
ENST00000440542.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
ENST00000440542.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
ENST00000443173.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
ENST00000443173.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
ENST00000443173.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
ENST00000443173.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
ENST00000443173.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
ENST00000443173.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
ENST00000443173.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
ENST00000443173.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
ENST00000454756.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
ENST00000454756.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
ENST00000454756.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
ENST00000454756.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
NM_001242690.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
NM_001242690.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
NM_001242690.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
NM_001242690.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
NM_001242690.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
NM_001242690.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
NM_001242690.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
NM_024534.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
NM_024534.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
NM_024534.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
NM_024534.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
NM_024534.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
NM_024534.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
NM_024534.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010694.hg.1 |
NM_024534.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400172833.hg.1 |
ENST00000440542.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400172833.hg.1 |
ENST00000443173.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400172833.hg.1 |
NM_001242690.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400172833.hg.1 |
NM_024534.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400172833.hg.1 |
ENST00000440542.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400172833.hg.1 |
ENST00000443173.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400172833.hg.1 |
NM_001242690.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400172833.hg.1 |
NM_024534.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400172833.hg.1 |
TC0400010694.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
219987_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
219987_at |
- |
- |
| AffymetrixPathwork Diagnostics Pathchip |
pw512591 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
219987_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
219987_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
219987_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
219987_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
219987_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
219987_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
219987_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
219987_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
219987_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
219987_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
219987_at5 |
- |
- |
| Human Genome U95 Set |
47831_at |
- |
- |
| Human Genome U95 Set |
47831_PM_at |
- |
- |