| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16000750.hg.1 |
ENST00000293897.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16000750.hg.1 |
ENST00000397547.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16000750.hg.1 |
ENST00000539762.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16000750.hg.1 |
NM_001053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16000750.hg.1 |
NM_001172560.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16000750.hg.1 |
TC16000035.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16000747.hg.1 |
ENST00000293897.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16000747.hg.1 |
NM_001053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16000747.hg.1 |
TC16000035.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16000746.hg.1 |
ENST00000397547.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16000746.hg.1 |
NM_001172560.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16000746.hg.1 |
TC16000035.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006519.hg.1 |
ENST00000293897.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006519.hg.1 |
ENST00000293897.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006519.hg.1 |
ENST00000293897.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006519.hg.1 |
ENST00000293897.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006519.hg.1 |
NM_001053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006519.hg.1 |
NM_001053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006519.hg.1 |
NM_001053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006519.hg.1 |
NM_001053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006519.hg.1 |
NM_001053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006519.hg.1 |
NM_001172560.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006519.hg.1 |
NM_001172560.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006519.hg.1 |
NM_001172560.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006519.hg.1 |
NM_001172560.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600006519.hg.1 |
NM_001172560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600145265.hg.1 |
ENST00000293897.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600145265.hg.1 |
NM_001053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600145265.hg.1 |
NM_001172560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600145265.hg.1 |
TC1600006519.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600145265.hg.1 |
ENST00000293897.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600145265.hg.1 |
NM_001053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600145265.hg.1 |
NM_001172560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006519.hg.1 |
ENST00000293897.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006519.hg.1 |
ENST00000293897.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006519.hg.1 |
ENST00000293897.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006519.hg.1 |
ENST00000293897.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600006519.hg.1 |
NM_001053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006519.hg.1 |
NM_001053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006519.hg.1 |
NM_001053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006519.hg.1 |
NM_001053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006519.hg.1 |
NM_001053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006519.hg.1 |
NM_001172560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006519.hg.1 |
NM_001172560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006519.hg.1 |
NM_001172560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006519.hg.1 |
NM_001172560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600006519.hg.1 |
NM_001172560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600145262.hg.1 |
ENST00000293897.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600145262.hg.1 |
NM_001053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600145262.hg.1 |
TC1600006519.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600145262.hg.1 |
ENST00000293897.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600145262.hg.1 |
NM_001053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600145261.hg.1 |
NM_001172560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600145261.hg.1 |
TC1600006519.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600145261.hg.1 |
NM_001172560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600145263.hg.1 |
ENST00000293897.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600145263.hg.1 |
NM_001053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600145263.hg.1 |
NM_001172560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600145263.hg.1 |
TC1600006519.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600145263.hg.1 |
ENST00000293897.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600145263.hg.1 |
NM_001053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600145263.hg.1 |
NM_001172560.1 |
Refseq |