Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00271279_s1
          See other PDHA2 GE Assays ›
          Gene Symbol:
          PDHA2
          Gene Name:
          pyruvate dehydrogenase (lipoamide) alpha 2
          Gene Aliases:
          PDHAL
          Chromosome Location:
          Chr.4: 95840088 - 95841474 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00271279_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PDHA2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005390.4 NP_005381.1 1 646 85
          GenBank mRNA AK313872.1 - 1 606 85
          BC030697.2 - 1 642 4824350 85
          BC066953.1 - 1 628 5273609 85
          BC094760.1 - 1 641 85
          BC119656.1 - 1 614 40115940 85
          BC119657.1 - 1 614 40115941 85
          BC127637.1 - 1 614 40115943 85
          BC127638.1 - 1 613 40115944 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0400008195.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC04000504.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PDHA2

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          glucose metabolic process
          acetyl-CoA biosynthetic process from pyruvate
          pyruvate metabolic process
          tricarboxylic acid cycle
          pyruvate dehydrogenase (acetyl-transferring) activity
          pyruvate dehydrogenase (NAD+) activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0400008195.hg.1 ENST00000295266.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400008195.hg.1 ENST00000295266.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400008195.hg.1 ENST00000295266.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400008195.hg.1 ENST00000295266.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400008195.hg.1 ENST00000295266.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400008195.hg.1 NM_005390.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400008195.hg.1 NM_005390.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400008195.hg.1 NM_005390.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400008195.hg.1 NM_005390.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04007427.hg.1 ENST00000295266.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04007427.hg.1 NM_005390.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04007427.hg.1 TC04000504.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0400008195.hg.1 ENST00000295266.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400008195.hg.1 ENST00000295266.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400008195.hg.1 ENST00000295266.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400008195.hg.1 ENST00000295266.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400008195.hg.1 ENST00000295266.5 Ensembl
          Affymetrix Human Clariom D Assay TC0400008195.hg.1 NM_005390.4 Refseq
          Affymetrix Human Clariom D Assay TC0400008195.hg.1 NM_005390.4 Refseq
          Affymetrix Human Clariom D Assay TC0400008195.hg.1 NM_005390.4 Refseq
          Affymetrix Human Clariom D Assay TC0400008195.hg.1 NM_005390.4 Refseq
          Affymetrix Human Clariom D Assay PSR0400156381.hg.1 ENST00000295266.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400156381.hg.1 NM_005390.4 Refseq
          Affymetrix Human Clariom D Assay PSR0400156381.hg.1 ENST00000295266.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400156381.hg.1 NM_005390.4 Refseq
          Affymetrix Human Clariom D Assay PSR0400156381.hg.1 TC0400008195.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400156382.hg.1 ENST00000295266.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400156382.hg.1 NM_005390.4 Refseq
          Affymetrix Human Clariom D Assay PSR0400156382.hg.1 ENST00000295266.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0400156382.hg.1 NM_005390.4 Refseq
          Affymetrix Human Clariom D Assay PSR0400156382.hg.1 TC0400008195.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 164213 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline