Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00287886_s1
          See other TMPO-AS1 GE Assays ›
          Gene Symbol:
          TMPO-AS1
          Gene Name:
          TMPO antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 98512973 - 98516226 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00287886_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TMPO-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027157.1 2 2016 89
          GenBank mRNA AK054586.1 - 1 648 89
          BC037346.2 - 1 1357 89

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 319476 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3210363 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_5054 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_5053 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 53742 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3210363 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3210363 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995789 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08562406 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995733 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995790 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995762 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995770 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08562359 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08562360 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995781 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08562390 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995732 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995792 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995767 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995771 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995731 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08562356 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995788 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995726 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995760 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995724 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995759 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995765 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995758 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995779 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995757 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995769 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995778 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995772 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995727 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08562350 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995783 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995786 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995773 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995730 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995774 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08562398 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995763 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995758 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995765 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08562350 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995770 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995733 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995779 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995727 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995789 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08562390 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08562398 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995726 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995783 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995730 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995792 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995759 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995769 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995774 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995773 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08562356 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995763 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995772 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995788 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995762 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995771 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08562359 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08562360 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995731 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995778 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08562406 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995732 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995760 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995786 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995781 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995767 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995757 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995790 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28995724 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P39786160 - -
          Agilent Human 1B Microarray G4111A A_32_P51951 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562332 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562406 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562409 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562345 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562385 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562355 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16584325 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562419 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562416 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562330 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562391 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562340 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562399 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562357 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562413 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562383 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562337 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562334 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562381 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562349 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562390 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562359 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562403 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562342 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16584313 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562388 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562401 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562394 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562378 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562405 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562353 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562396 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562339 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562347 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16584323 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08562343 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 227578_PM_at - -
          Human Genome U133 Plus 2.0 Array 227578_PM_at - -
          Human Genome U133 Plus 2.0 Array 227578_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227578_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227578_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227578_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227578_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227578_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227578_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227578_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227578_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227578_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227578_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227578_at11 - -
          Human Genome U95 Set 45702_at - -
          Human Genome U95 Set 45702_PM_at - -
          Human Genome U133 Set 227578_PM_at - -
          Human Genome U133 Set 227578_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A047924 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A047924 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3266964 NR_027157.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3266964 AK054586.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3176746 BC037346.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3176746 NR_027157.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3176746 AK054586.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline