Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00292485_m1
          See other DNAH5 GE Assays ›
          Gene Symbol:
          DNAH5
          Gene Name:
          dynein axonemal heavy chain 5
          Gene Aliases:
          CILD3, DNAHC5, HL1, KTGNR, PCD
          Chromosome Location:
          Chr.5: 13690328 - 14011829 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00292485_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DNAH5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001369.2 NP_001360.1 42 - 43 7030 74
          XM_005248262.3 XP_005248319.2 42 - 43 7113 74
          XM_017009177.1 XP_016864666.1 42 - 43 7113 74
          XM_017009178.1 XP_016864667.1 42 - 43 7916 74
          XM_017009179.1 XP_016864668.1 36 - 37 6210 74
          XM_017009180.1 XP_016864669.1 42 - 43 7113 74
          XM_017009181.1 XP_016864670.1 42 - 43 7113 74
          XM_017009182.1 XP_016864671.1 42 - 43 7113 74
          XM_017009183.1 XP_016864672.1 42 - 43 7113 74
          XM_017009184.1 XP_016864673.1 42 - 43 7113 74
          XM_017009185.1 XP_016864674.1 14 - 15 2409 74
          XM_017009186.1 XP_016864675.1 11 - 12 1825 74
          XM_017009187.1 XP_016864676.1 42 - 43 7113 74
          XM_017009188.1 XP_016864677.1 8 - 9 2641 74
          GenBank mRNA AY045575.1 - 42 - 43 7030 74
          AY049075.1 - 7 - 8 1123 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500010148.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC05001171.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene DNAH5

          Panther Classification:

          Molecular Function -

          microtubule binding motor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cilium movement
          determination of left/right symmetry
          heart development
          biological_process
          lateral ventricle development
          sperm motility
          outer dynein arm assembly
          cilium assembly
          microtubule motor activity
          ATP binding
          ATPase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR05016749.hg.1 ENST00000265104.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05016749.hg.1 NM_001369.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05016749.hg.1 TC05001171.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0500171077.hg.1 ENST00000265104.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0500171077.hg.1 NM_001369.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500171077.hg.1 ENST00000265104.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0500171077.hg.1 NM_001369.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500171077.hg.1 TC0500010148.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline