Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00293264_m1
          See other ST6GAL2 GE Assays ›
          Gene Symbol:
          ST6GAL2
          Gene Name:
          ST6 beta-galactoside alpha-2,6-sialyltransferase 2
          Gene Aliases:
          SIAT2, ST6GalII
          Chromosome Location:
          Chr.2: 106801600 - 106887410 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00293264_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ST6GAL2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001142352.1 NP_001135824.1 1 - 2 136 125
          NM_032528.2 NP_115917.1 1 - 2 136 125
          XM_017005110.1 XP_016860599.1 1 - 2 87 125
          GenBank mRNA AB059555.1 - 1 - 2 122 124
          AK307784.1 - 1 - 2 137 125
          BC008680.1 - 1 - 2 122 3856171 124
          DA176100.1 - 1 - 2 137 125

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200013779.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02002174.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AB059555.1,BC008680.1,NM_001142352.1,NM_032528.2,XM_017005110.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ST6GAL2

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          oligosaccharide metabolic process
          protein N-linked glycosylation via asparagine
          growth
          sialylation
          beta-galactoside alpha-2,6-sialyltransferase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02035396.hg.1 ENST00000361686.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02035396.hg.1 ENST00000409087.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02035396.hg.1 ENST00000409382.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02035396.hg.1 NM_001142351.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02035396.hg.1 NM_001142352.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02035396.hg.1 NM_032528.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02035396.hg.1 TC02002174.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000361686.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000361686.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000361686.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000361686.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000361686.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000361686.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000361686.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000361686.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000361686.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000361686.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000361686.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000361803.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000361803.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000361803.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000409087.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000409087.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000409087.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000409087.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000409087.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000409087.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000409087.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000409087.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000409087.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000409382.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000409382.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000409382.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000409382.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000409382.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000409382.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000409382.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000409382.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000409382.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000409382.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000409382.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000409382.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000419159.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000419159.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 ENST00000419159.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_001142351.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_001142351.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_001142351.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_001142351.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_001142351.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_001142351.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_001142351.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_001142351.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_001142351.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_001142351.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_001142351.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_001142352.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_001142352.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_001142352.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_001142352.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_001142352.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_001142352.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_001142352.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_001142352.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_001142352.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_032528.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_032528.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_032528.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_032528.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_032528.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_032528.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_032528.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_032528.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_032528.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_032528.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200013779.hg.1 NM_032528.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02035400.hg.1 ENST00000361686.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02035400.hg.1 ENST00000409087.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02035400.hg.1 NM_001142352.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02035400.hg.1 NM_032528.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02035400.hg.1 TC02002174.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02035402.hg.1 ENST00000361686.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02035402.hg.1 ENST00000409087.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02035402.hg.1 NM_001142352.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02035402.hg.1 NM_032528.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02035402.hg.1 TC02002174.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000361686.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000361686.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000361686.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000361686.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000361686.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000361686.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000361686.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000361686.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000361686.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000361686.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000361686.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000361803.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000361803.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000361803.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000409087.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000409087.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000409087.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000409087.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000409087.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000409087.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000409087.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000409087.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000409087.3 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000409382.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000409382.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000409382.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000409382.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000409382.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000409382.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000409382.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000409382.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000409382.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000409382.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000409382.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000409382.7 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000419159.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000419159.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 ENST00000419159.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_001142351.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_001142351.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_001142351.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_001142351.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_001142351.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_001142351.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_001142351.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_001142351.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_001142351.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_001142351.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_001142351.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_001142352.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_001142352.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_001142352.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_001142352.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_001142352.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_001142352.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_001142352.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_001142352.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_001142352.1 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_032528.2 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_032528.2 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_032528.2 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_032528.2 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_032528.2 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_032528.2 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_032528.2 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_032528.2 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_032528.2 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_032528.2 Refseq
          Affymetrix Human Clariom D Assay TC0200013779.hg.1 NM_032528.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200201034.hg.1 ENST00000361686.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201034.hg.1 ENST00000409087.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201034.hg.1 NM_001142352.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200201034.hg.1 NM_032528.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200201034.hg.1 ENST00000361686.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201034.hg.1 ENST00000409087.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201034.hg.1 NM_001142352.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200201034.hg.1 NM_032528.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200201034.hg.1 TC0200013779.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200201030.hg.1 ENST00000361686.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201030.hg.1 ENST00000409087.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201030.hg.1 ENST00000409382.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201030.hg.1 ENST00000419159.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201030.hg.1 NM_001142351.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200201030.hg.1 NM_001142352.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200201030.hg.1 NM_032528.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200201030.hg.1 ENST00000361686.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201030.hg.1 ENST00000409087.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201030.hg.1 ENST00000409382.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201030.hg.1 ENST00000419159.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200201030.hg.1 NM_001142351.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200201030.hg.1 NM_001142352.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200201030.hg.1 NM_032528.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200201030.hg.1 TC0200013779.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 216870 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline