Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00299694_m1
          See other ANKS1A GE Assays ›
          Gene Symbol:
          ANKS1A
          Gene Name:
          ankyrin repeat and sterile alpha motif domain containing 1A
          Gene Aliases:
          ANKS1
          Chromosome Location:
          Chr.6: 34889261 - 35097994 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00299694_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ANKS1A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_015245.2 NP_056060.2 8 - 9 1352 72
          XM_005248964.3 XP_005249021.1 9 - 10 1403 72
          XM_006715036.2 XP_006715099.1 9 - 10 1403 72
          XM_011514431.2 XP_011512733.1 9 - 10 1403 72
          XM_011514432.2 XP_011512734.1 9 - 10 1403 72
          XM_011514433.1 XP_011512735.1 9 - 10 1403 72
          XM_011514434.2 XP_011512736.1 8 - 9 1340 72
          XM_011514435.2 XP_011512737.1 8 - 9 1340 72
          XM_011514436.2 XP_011512738.1 9 - 10 4972 72
          GenBank mRNA BC022396.1 - 8 - 9 1352 72
          BC031934.1 - 9 - 10 1424 4819948 72
          BC132832.1 - 8 - 9 1289 40146854 72
          D86982.1 - 8 - 9 1349 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600007774.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06000510.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the BC031934.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ANKS1A

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          substrate-dependent cell migration
          neuron remodeling
          ephrin receptor signaling pathway
          regulation of ephrin receptor signaling pathway
          protein binding
          ephrin receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR06005361.hg.1 ENST00000360359.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06005361.hg.1 ENST00000373990.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06005361.hg.1 ENST00000544150.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06005361.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06005361.hg.1 TC06000510.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06005359.hg.1 ENST00000360359.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06005359.hg.1 ENST00000373990.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06005359.hg.1 ENST00000544150.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06005359.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06005359.hg.1 TC06000510.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000470698.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000470698.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000470698.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000470698.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000470698.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 ENST00000470698.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06005360.hg.1 ENST00000360359.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06005360.hg.1 ENST00000373990.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06005360.hg.1 ENST00000544150.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06005360.hg.1 NM_015245.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06005360.hg.1 TC06000510.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0600153885.hg.1 TC0600007774.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000470698.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000470698.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000470698.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000470698.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000470698.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 ENST00000470698.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007774.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600153882.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0600153882.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600153882.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0600153882.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600153882.hg.1 TC0600007774.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600153883.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0600153883.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600153883.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0600153883.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600153883.hg.1 TC0600007774.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600153881.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0600153881.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600153881.hg.1 ENST00000360359.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0600153881.hg.1 NM_015245.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600153881.hg.1 TC0600007774.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600153886.hg.1 TC0600007774.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline