| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21000814.hg.1 |
ENST00000333337.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21000814.hg.1 |
ENST00000382357.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21000814.hg.1 |
NM_005806.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21000814.hg.1 |
TC21000123.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006948.hg.1 |
ENST00000333337.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006948.hg.1 |
ENST00000333337.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006948.hg.1 |
ENST00000333337.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006948.hg.1 |
ENST00000333337.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006948.hg.1 |
ENST00000333337.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006948.hg.1 |
ENST00000333337.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006948.hg.1 |
ENST00000333337.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006948.hg.1 |
ENST00000382357.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006948.hg.1 |
ENST00000382357.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006948.hg.1 |
ENST00000382357.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006948.hg.1 |
ENST00000382357.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006948.hg.1 |
ENST00000382357.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006948.hg.1 |
ENST00000382357.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006948.hg.1 |
ENST00000382357.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006948.hg.1 |
ENST00000430860.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006948.hg.1 |
ENST00000430860.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006948.hg.1 |
ENST00000430860.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006948.hg.1 |
ENST00000430860.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006948.hg.1 |
NM_005806.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006948.hg.1 |
NM_005806.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006948.hg.1 |
NM_005806.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006948.hg.1 |
NM_005806.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006948.hg.1 |
NM_005806.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006948.hg.1 |
NM_005806.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21000813.hg.1 |
ENST00000333337.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21000813.hg.1 |
ENST00000382357.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21000813.hg.1 |
NM_005806.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21000813.hg.1 |
TC21000123.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR2100146618.hg.1 |
ENST00000333337.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100146618.hg.1 |
ENST00000382357.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100146618.hg.1 |
NM_005806.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100146618.hg.1 |
TC2100006948.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100146618.hg.1 |
ENST00000333337.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100146618.hg.1 |
ENST00000382357.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100146618.hg.1 |
NM_005806.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100146619.hg.1 |
ENST00000333337.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100146619.hg.1 |
ENST00000382357.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100146619.hg.1 |
NM_005806.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100146619.hg.1 |
TC2100006948.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100146619.hg.1 |
ENST00000333337.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100146619.hg.1 |
ENST00000382357.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100146619.hg.1 |
NM_005806.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100146615.hg.1 |
ENST00000333337.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100146615.hg.1 |
TC2100006948.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100146615.hg.1 |
ENST00000333337.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006948.hg.1 |
ENST00000333337.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006948.hg.1 |
ENST00000333337.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006948.hg.1 |
ENST00000333337.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006948.hg.1 |
ENST00000333337.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006948.hg.1 |
ENST00000333337.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006948.hg.1 |
ENST00000333337.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006948.hg.1 |
ENST00000333337.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006948.hg.1 |
ENST00000382357.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006948.hg.1 |
ENST00000382357.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006948.hg.1 |
ENST00000382357.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006948.hg.1 |
ENST00000382357.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006948.hg.1 |
ENST00000382357.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006948.hg.1 |
ENST00000382357.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006948.hg.1 |
ENST00000382357.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006948.hg.1 |
ENST00000430860.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006948.hg.1 |
ENST00000430860.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006948.hg.1 |
ENST00000430860.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006948.hg.1 |
ENST00000430860.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006948.hg.1 |
NM_005806.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006948.hg.1 |
NM_005806.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006948.hg.1 |
NM_005806.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006948.hg.1 |
NM_005806.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006948.hg.1 |
NM_005806.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006948.hg.1 |
NM_005806.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100146614.hg.1 |
ENST00000333337.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100146614.hg.1 |
ENST00000430860.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100146614.hg.1 |
TC2100006948.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100146614.hg.1 |
ENST00000333337.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100146614.hg.1 |
ENST00000430860.1 |
Ensembl |