| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18004001.hg.1 |
ENST00000327606.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18004001.hg.1 |
NM_000529.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18004001.hg.1 |
TC18000382.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18004006.hg.1 |
ENST00000327606.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18004006.hg.1 |
NM_000529.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18004006.hg.1 |
TC18000382.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18004002.hg.1 |
ENST00000327606.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18004002.hg.1 |
NM_000529.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18004002.hg.1 |
TC18000382.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18004004.hg.1 |
ENST00000327606.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18004004.hg.1 |
NM_000529.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18004004.hg.1 |
TC18000382.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009098.hg.1 |
ENST00000390765.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008166.hg.1 |
ENST00000327606.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008166.hg.1 |
ENST00000327606.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008166.hg.1 |
ENST00000327606.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008166.hg.1 |
ENST00000327606.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008166.hg.1 |
ENST00000327606.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008166.hg.1 |
ENST00000327606.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008166.hg.1 |
ENST00000399821.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008166.hg.1 |
ENST00000399821.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008166.hg.1 |
ENST00000399821.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008166.hg.1 |
NM_000529.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008166.hg.1 |
NM_000529.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008166.hg.1 |
NM_000529.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008166.hg.1 |
NM_000529.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008166.hg.1 |
NM_000529.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008166.hg.1 |
NM_000529.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008166.hg.1 |
NM_001291911.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008166.hg.1 |
NM_001291911.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008166.hg.1 |
NM_001291911.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008166.hg.1 |
NM_001291911.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008166.hg.1 |
NM_001291911.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008166.hg.1 |
NM_001291911.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800155817.hg.1 |
ENST00000327606.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155817.hg.1 |
NM_000529.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800155817.hg.1 |
NM_001291911.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800155817.hg.1 |
ENST00000327606.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155817.hg.1 |
NM_000529.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800155817.hg.1 |
NM_001291911.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800155817.hg.1 |
TC1800008166.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1300009098.hg.1 |
ENST00000390765.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008166.hg.1 |
ENST00000327606.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008166.hg.1 |
ENST00000327606.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008166.hg.1 |
ENST00000327606.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008166.hg.1 |
ENST00000327606.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008166.hg.1 |
ENST00000327606.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008166.hg.1 |
ENST00000327606.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008166.hg.1 |
ENST00000399821.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008166.hg.1 |
ENST00000399821.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008166.hg.1 |
ENST00000399821.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008166.hg.1 |
NM_000529.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008166.hg.1 |
NM_000529.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008166.hg.1 |
NM_000529.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008166.hg.1 |
NM_000529.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008166.hg.1 |
NM_000529.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008166.hg.1 |
NM_000529.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008166.hg.1 |
NM_001291911.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008166.hg.1 |
NM_001291911.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008166.hg.1 |
NM_001291911.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008166.hg.1 |
NM_001291911.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008166.hg.1 |
NM_001291911.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008166.hg.1 |
NM_001291911.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800155816.hg.1 |
ENST00000327606.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155816.hg.1 |
NM_000529.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800155816.hg.1 |
NM_001291911.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800155816.hg.1 |
ENST00000327606.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800155816.hg.1 |
NM_000529.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800155816.hg.1 |
NM_001291911.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800155816.hg.1 |
TC1800008166.hg.1 |
Clariom D TC |